Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47412285-47412444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH2
Entrez Gene ID
4436
Gene Name
mutS homolog 2
Gene Aliases
COCA1, FCC1, HNPCC, HNPCC1, LCFS2
UniGene
Hs.597656
Gene Chromosome Location
Chr.2: 47403067-47634501 on Build GRCh38
Transcripts
NM_000251.2 NM_001258281.1 XM_005264332.3 XM_011532867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5465572 c.646-1G>T p.? Chr.2: 47412413-47412413 on Build GRCh38
26121 c.658G>T p.G220* Chr.2: 47412426-47412426 on Build GRCh38
4915799 c.646-2A>T p.? Chr.2: 47412412-47412412 on Build GRCh38
327050 c.662G>T p.G221V Chr.2: 47412430-47412430 on Build GRCh38
6976564 c.667_706del40 p.L223fs*10 Chr.2: 47412435-47412474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781033756 Chr.2: 47412363-47412363 on Build GRCh38
rs752468406 Chr.2: 47412368-47412368 on Build GRCh38
rs377432684 Chr.2: 47412321-47412321 on Build GRCh38
rs1027167224 Chr.2: 47412385-47412385 on Build GRCh38
rs749322734 Chr.2: 47412382-47412382 on Build GRCh38
rs63749936 Chr.2: 47412414-47412414 on Build GRCh38
rs879254124 Chr.2: 47412403-47412403 on Build GRCh38
rs780285303 Chr.2: 47412374-47412374 on Build GRCh38
rs769955966 Chr.2: 47412342-47412342 on Build GRCh38
rs140723233 Chr.2: 47412396-47412396 on Build GRCh38
rs774623661 Chr.2: 47412391-47412391 on Build GRCh38
rs761205332 Chr.2: 47412401-47412401 on Build GRCh38
rs267607929 Chr.2: 47412411-47412422 on Build GRCh38
rs1004540142 Chr.2: 47412347-47412347 on Build GRCh38
rs587779169 Chr.2: 47412412-47412412 on Build GRCh38
rs908088330 Chr.2: 47412339-47412339 on Build GRCh38
rs587779974 Chr.2: 47412410-47412410 on Build GRCh38
rs182754158 Chr.2: 47412372-47412372 on Build GRCh38
rs576478719 Chr.2: 47412383-47412383 on Build GRCh38
rs63751602 Chr.2: 47412418-47412422 on Build GRCh38
rs759385803 Chr.2: 47412383-47412385 on Build GRCh38
rs565117135 Chr.2: 47412398-47412398 on Build GRCh38
rs267607930 Chr.2: 47412411-47412411 on Build GRCh38
rs762436663 Chr.2: 47412417-47412417 on Build GRCh38
rs786203108 Chr.2: 47412415-47412415 on Build GRCh38
rs773520113 Chr.2: 47412399-47412399 on Build GRCh38
rs878853820 Chr.2: 47412406-47412406 on Build GRCh38

View Full Product Details