Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68810135-68810270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH1
Entrez Gene ID
999
Gene Name
cadherin 1
Gene Aliases
Arc-1, CD324, CDHE, ECAD, LCAM, UVO
UniGene
Hs.461086
Gene Chromosome Location
Chr.16: 68737290-68835542 on Build GRCh38
Transcripts
NM_001317184.1 NM_001317185.1 NM_001317186.1 NM_004360.4 XM_011523488.1 XM_011523489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
35882 c.688_832del145 p.L230fs*4 Chr.16: 68810197-68810341 on Build GRCh38
6910322 c.739G>T p.E247* Chr.16: 68810248-68810248 on Build GRCh38
6400615 c.752C>T p.T251M Chr.16: 68810261-68810261 on Build GRCh38
19516 c.760G>T p.D254Y Chr.16: 68810269-68810269 on Build GRCh38
19419 c.727G>T p.E243* Chr.16: 68810236-68810236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774408716 Chr.16: 68810241-68810241 on Build GRCh38
rs371612417 Chr.16: 68810243-68810243 on Build GRCh38
rs111662525 Chr.16: 68810233-68810233 on Build GRCh38
rs752284537 Chr.16: 68810158-68810158 on Build GRCh38
rs587780788 Chr.16: 68810228-68810228 on Build GRCh38
rs587780537 Chr.16: 68810224-68810224 on Build GRCh38
rs189969617 Chr.16: 68810183-68810183 on Build GRCh38
rs1050660802 Chr.16: 68810187-68810187 on Build GRCh38
rs115494727 Chr.16: 68810208-68810208 on Build GRCh38
rs781633588 Chr.16: 68810193-68810193 on Build GRCh38
rs747621972 Chr.16: 68810172-68810172 on Build GRCh38
rs878854694 Chr.16: 68810209-68810209 on Build GRCh38
rs1014221041 Chr.16: 68810176-68810176 on Build GRCh38
rs730881664 Chr.16: 68810221-68810221 on Build GRCh38
rs780297063 Chr.16: 68810223-68810223 on Build GRCh38
rs863224397 Chr.16: 68810199-68810199 on Build GRCh38
rs768781036 Chr.16: 68810232-68810232 on Build GRCh38
rs863224398 Chr.16: 68810217-68810217 on Build GRCh38
rs730881663 Chr.16: 68810216-68810216 on Build GRCh38
rs758019856 Chr.16: 68810161-68810161 on Build GRCh38
rs749491453 Chr.16: 68810226-68810226 on Build GRCh38
rs777412759 Chr.16: 68810169-68810169 on Build GRCh38
rs892721831 Chr.16: 68810166-68810166 on Build GRCh38

View Full Product Details