Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73998393-73998529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PML
Entrez Gene ID
5371
Gene Name
promyelocytic leukemia
Gene Aliases
MYL, PP8675, RNF71, TRIM19
UniGene
Hs.526464
Gene Chromosome Location
Chr.15: 73994673-74047819 on Build GRCh38
Transcripts
NM_002675.3 NM_033238.2 NM_033239.2 NM_033240.2 NM_033244.3 NM_033246.2 NM_033247.2 NM_033249.2 NM_033250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1236512 c.564C>A p.F188L Chr.15: 73998438-73998438 on Build GRCh38
5029375 c.556A>G p.N186D Chr.15: 73998430-73998430 on Build GRCh38
4454684 c.557A>G p.N186S Chr.15: 73998431-73998431 on Build GRCh38
4454685 c.557A>G p.N186S Chr.15: 73998431-73998431 on Build GRCh38
5029376 c.556A>G p.N186D Chr.15: 73998430-73998430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566690133 Chr.15: 73998481-73998481 on Build GRCh38
rs200143816 Chr.15: 73998429-73998429 on Build GRCh38
rs754423752 Chr.15: 73998506-73998506 on Build GRCh38
rs750773492 Chr.15: 73998484-73998484 on Build GRCh38
rs771660433 Chr.15: 73998418-73998418 on Build GRCh38
rs996506100 Chr.15: 73998431-73998431 on Build GRCh38
rs780683954 Chr.15: 73998495-73998495 on Build GRCh38
rs879233597 Chr.15: 73998494-73998494 on Build GRCh38
rs138345086 Chr.15: 73998443-73998443 on Build GRCh38
rs775451143 Chr.15: 73998452-73998452 on Build GRCh38
rs780737061 Chr.15: 73998411-73998411 on Build GRCh38
rs373742710 Chr.15: 73998436-73998436 on Build GRCh38
rs756585678 Chr.15: 73998487-73998487 on Build GRCh38
rs35291318 Chr.15: 73998461-73998461 on Build GRCh38
rs762034131 Chr.15: 73998479-73998479 on Build GRCh38
rs751591785 Chr.15: 73998462-73998462 on Build GRCh38
rs746745053 Chr.15: 73998426-73998426 on Build GRCh38
rs759349668 Chr.15: 73998446-73998446 on Build GRCh38
rs747580265 Chr.15: 73998414-73998414 on Build GRCh38
rs551261745 Chr.15: 73998451-73998451 on Build GRCh38
rs367778787 Chr.15: 73998456-73998456 on Build GRCh38

View Full Product Details