Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42129112-42129255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2D6
Entrez Gene ID
1565
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
UniGene
Hs.648256
Gene Chromosome Location
Chr.22: 42125531-42130881 on Build GRCh38
Transcripts
NM_000106.5 NM_001025161.2 XM_011529966.2 XM_011529968.2 XM_011529970.2 XM_011529972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3785680 c.367C>A p.R123S Chr.22: 42129171-42129171 on Build GRCh38
3061699 c.358T>A p.F120I Chr.22: 42129180-42129180 on Build GRCh38
4430497 c.367C>T p.R123C Chr.22: 42129171-42129171 on Build GRCh38
6162114 c.410C>T p.S137F Chr.22: 42129128-42129128 on Build GRCh38
6448916 c.393G>C p.Q131H Chr.22: 42129145-42129145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374852685 Chr.22: 42129168-42129168 on Build GRCh38
rs61736512 Chr.22: 42129132-42129132 on Build GRCh38
rs369043970 Chr.22: 42129160-42129160 on Build GRCh38
rs1135822 Chr.22: 42129180-42129180 on Build GRCh38
rs373668214 Chr.22: 42129228-42129228 on Build GRCh38
rs377591409 Chr.22: 42129173-42129173 on Build GRCh38
rs745756780 Chr.22: 42129211-42129211 on Build GRCh38
rs267608303 Chr.22: 42129171-42129171 on Build GRCh38
rs753856865 Chr.22: 42129196-42129196 on Build GRCh38
rs750894050 Chr.22: 42129201-42129201 on Build GRCh38
rs753911276 Chr.22: 42129186-42129187 on Build GRCh38
rs1053431848 Chr.22: 42129146-42129146 on Build GRCh38
rs773878068 Chr.22: 42129210-42129210 on Build GRCh38
rs768749385 Chr.22: 42129216-42129216 on Build GRCh38
rs376636053 Chr.22: 42129156-42129156 on Build GRCh38
rs777909589 Chr.22: 42129148-42129148 on Build GRCh38
rs772046082 Chr.22: 42129212-42129212 on Build GRCh38
rs755309061 Chr.22: 42129234-42129234 on Build GRCh38
rs374616348 Chr.22: 42129183-42129183 on Build GRCh38
rs765422829 Chr.22: 42129199-42129199 on Build GRCh38
rs775232205 Chr.22: 42129213-42129213 on Build GRCh38
rs769513307 Chr.22: 42129181-42129181 on Build GRCh38
rs370267861 Chr.22: 42129225-42129225 on Build GRCh38
rs757417430 Chr.22: 42129198-42129198 on Build GRCh38
rs761946374 Chr.22: 42129224-42129224 on Build GRCh38
rs1011963265 Chr.22: 42129153-42129153 on Build GRCh38
rs377440666 Chr.22: 42129202-42129202 on Build GRCh38
rs761571009 Chr.22: 42129157-42129157 on Build GRCh38
rs550056038 Chr.22: 42129133-42129133 on Build GRCh38
rs267608304 Chr.22: 42129184-42129184 on Build GRCh38
rs751868209 Chr.22: 42129233-42129233 on Build GRCh38
rs767035201 Chr.22: 42129231-42129231 on Build GRCh38
rs771490928 Chr.22: 42129150-42129150 on Build GRCh38
rs1135823 Chr.22: 42129174-42129174 on Build GRCh38
rs747460472 Chr.22: 42129204-42129204 on Build GRCh38
rs777551180 Chr.22: 42129208-42129208 on Build GRCh38
rs562633069 Chr.22: 42129191-42129191 on Build GRCh38
rs779123374 Chr.22: 42129151-42129151 on Build GRCh38
rs76060075 Chr.22: 42129185-42129185 on Build GRCh38
rs80018788 Chr.22: 42129162-42129162 on Build GRCh38
rs766391487 Chr.22: 42129166-42129166 on Build GRCh38
rs61736507 Chr.22: 42129178-42129178 on Build GRCh38
rs759470167 Chr.22: 42129188-42129188 on Build GRCh38
rs756086368 Chr.22: 42129145-42129145 on Build GRCh38
rs781457579 Chr.22: 42129134-42129134 on Build GRCh38
rs772813082 Chr.22: 42129159-42129159 on Build GRCh38
rs766505316 Chr.22: 42129178-42129179 on Build GRCh38
rs767255540 Chr.22: 42129190-42129190 on Build GRCh38
rs770357599 Chr.22: 42129209-42129209 on Build GRCh38
rs373000587 Chr.22: 42129170-42129170 on Build GRCh38
rs574423474 Chr.22: 42129214-42129214 on Build GRCh38
rs1058164 Chr.22: 42129130-42129130 on Build GRCh38
rs752758046 Chr.22: 42129143-42129143 on Build GRCh38
rs528764348 Chr.22: 42129237-42129237 on Build GRCh38
rs746272785 Chr.22: 42129152-42129152 on Build GRCh38
rs867339178 Chr.22: 42129136-42129136 on Build GRCh38
rs756389811 Chr.22: 42129172-42129172 on Build GRCh38
rs267608305 Chr.22: 42129193-42129193 on Build GRCh38

View Full Product Details