Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3150900-3151054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

Gene Symbol
TRNT1
Entrez Gene ID
51095
Gene Name
tRNA nucleotidyl transferase 1
Gene Aliases
CCA1, CGI-47, MtCCA, RPEM, SIFD
UniGene
Hs.732725
Gene Chromosome Location
Chr.3: 3126916-3153435 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883813 c.1271C>A p.P424H Chr.3: 3150923-3150923 on Build GRCh38
1043611 c.1183G>A p.A395T Chr.3: 3151011-3151011 on Build GRCh38
6999639 c.1294G>A p.E432K Chr.3: 3150900-3150900 on Build GRCh38
1043612 c.1178T>C p.I393T Chr.3: 3151016-3151016 on Build GRCh38
4116869 c.1255C>T p.R419* Chr.3: 3150939-3150939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139391260 Chr.3: 3150974-3150974 on Build GRCh38
rs959958369 Chr.3: 3150979-3150979 on Build GRCh38
rs750260374 Chr.3: 3151011-3151011 on Build GRCh38
rs763710763 Chr.3: 3151000-3151000 on Build GRCh38
rs751561359 Chr.3: 3151007-3151007 on Build GRCh38
rs748311975 Chr.3: 3151029-3151029 on Build GRCh38
rs116847323 Chr.3: 3150985-3150985 on Build GRCh38
rs761596274 Chr.3: 3151002-3151002 on Build GRCh38
rs760208270 Chr.3: 3150986-3150986 on Build GRCh38
rs755083658 Chr.3: 3151018-3151018 on Build GRCh38
rs751645075 Chr.3: 3151017-3151017 on Build GRCh38
rs121918368 Chr.3: 3150939-3150939 on Build GRCh38
rs758347001 Chr.3: 3151012-3151012 on Build GRCh38
rs781369096 Chr.3: 3151026-3151026 on Build GRCh38
rs760799393 Chr.3: 3150938-3150938 on Build GRCh38
rs753401262 Chr.3: 3151001-3151001 on Build GRCh38
rs201809570 Chr.3: 3151010-3151010 on Build GRCh38
rs142752214 Chr.3: 3150970-3150970 on Build GRCh38
rs1023603838 Chr.3: 3151024-3151024 on Build GRCh38
rs766454644 Chr.3: 3151013-3151013 on Build GRCh38
rs199720401 Chr.3: 3150952-3150952 on Build GRCh38
rs771439054 Chr.3: 3150977-3150977 on Build GRCh38
rs200533689 Chr.3: 3150976-3150976 on Build GRCh38
rs770018396 Chr.3: 3150972-3150972 on Build GRCh38
rs797045036 Chr.3: 3151023-3151023 on Build GRCh38
rs781294762 Chr.3: 3150966-3150966 on Build GRCh38
rs200513126 Chr.3: 3150940-3150940 on Build GRCh38

View Full Product Details