Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33319669-33319840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAXX
Entrez Gene ID
1616
Gene Name
death domain associated protein
Gene Aliases
BING2, DAP6, EAP1
UniGene
Hs.336916
Gene Chromosome Location
Chr.6: 33318558-33323016 on Build GRCh38
Transcripts
NM_001141969.1 NM_001141970.1 NM_001254717.1 NM_001350.4 XM_005248860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3873168 c.1577G>A p.R526H Chr.6: 33319743-33319743 on Build GRCh38
5357385 c.1486C>A p.P496T Chr.6: 33319834-33319834 on Build GRCh38
6921331 c.1595T>A p.L532* Chr.6: 33319725-33319725 on Build GRCh38
7199027 c.1530T>C p.P510P Chr.6: 33319790-33319790 on Build GRCh38
6982595 c.1612delC p.L538fs*6 Chr.6: 33319708-33319708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141395832 Chr.6: 33319743-33319743 on Build GRCh38
rs760674231 Chr.6: 33319788-33319788 on Build GRCh38
rs773386948 Chr.6: 33319710-33319710 on Build GRCh38
rs61748592 Chr.6: 33319727-33319727 on Build GRCh38
rs773879056 Chr.6: 33319744-33319744 on Build GRCh38
rs772967665 Chr.6: 33319770-33319770 on Build GRCh38
rs753718788 Chr.6: 33319793-33319793 on Build GRCh38
rs146868243 Chr.6: 33319809-33319809 on Build GRCh38
rs758413375 Chr.6: 33319802-33319802 on Build GRCh38
rs761486064 Chr.6: 33319755-33319755 on Build GRCh38
rs775204775 Chr.6: 33319728-33319728 on Build GRCh38
rs748790861 Chr.6: 33319735-33319735 on Build GRCh38
rs766292072 Chr.6: 33319792-33319792 on Build GRCh38
rs372876513 Chr.6: 33319750-33319750 on Build GRCh38
rs764801966 Chr.6: 33319728-33319733 on Build GRCh38
rs767236551 Chr.6: 33319763-33319763 on Build GRCh38
rs778573009 Chr.6: 33319785-33319785 on Build GRCh38
rs755821596 Chr.6: 33319808-33319808 on Build GRCh38

View Full Product Details