Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3119174-3119336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971905 c.767G>A p.R256Q Chr.19: 3119237-3119237 on Build GRCh38
6672108 c.759C>A p.A253A Chr.19: 3119229-3119229 on Build GRCh38
6963364 c.790T>C p.F264L Chr.19: 3119260-3119260 on Build GRCh38
6229418 c.804C>T p.S268S Chr.19: 3119274-3119274 on Build GRCh38
2750397 c.736-8C>T p.? Chr.19: 3119198-3119198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369692834 Chr.19: 3119199-3119199 on Build GRCh38
rs775040372 Chr.19: 3119236-3119236 on Build GRCh38
rs750681091 Chr.19: 3119274-3119274 on Build GRCh38
rs544105544 Chr.19: 3119194-3119194 on Build GRCh38
rs377219320 Chr.19: 3119198-3119198 on Build GRCh38
rs761542744 Chr.19: 3119248-3119248 on Build GRCh38
rs776428312 Chr.19: 3119244-3119244 on Build GRCh38
rs771537617 Chr.19: 3119232-3119232 on Build GRCh38
rs745528917 Chr.19: 3119220-3119220 on Build GRCh38
rs529942770 Chr.19: 3119237-3119237 on Build GRCh38
rs144829732 Chr.19: 3119275-3119275 on Build GRCh38
rs751628153 Chr.19: 3119283-3119283 on Build GRCh38
rs116848044 Chr.19: 3119200-3119200 on Build GRCh38
rs997103699 Chr.19: 3119230-3119230 on Build GRCh38
rs755573572 Chr.19: 3119286-3119286 on Build GRCh38
rs765454337 Chr.19: 3119259-3119259 on Build GRCh38
rs770720811 Chr.19: 3119201-3119201 on Build GRCh38
rs4900 Chr.19: 3119241-3119241 on Build GRCh38
rs755520418 Chr.19: 3119195-3119195 on Build GRCh38
rs778362669 Chr.19: 3119209-3119209 on Build GRCh38

View Full Product Details