Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3110067-3110230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433596 c.155A>G p.K52R Chr.19: 3110167-3110167 on Build GRCh38
4827188 c.168C>G p.I56M Chr.19: 3110180-3110180 on Build GRCh38
4076638 c.169A>C p.K57Q Chr.19: 3110181-3110181 on Build GRCh38
178570 c.196G>A p.G66S Chr.19: 3110208-3110208 on Build GRCh38
5783069 c.166A>T p.I56F Chr.19: 3110178-3110178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762506413 Chr.19: 3110104-3110104 on Build GRCh38
rs368986120 Chr.19: 3110124-3110124 on Build GRCh38
rs61731117 Chr.19: 3110153-3110153 on Build GRCh38
rs772594536 Chr.19: 3110138-3110138 on Build GRCh38
rs7255425 Chr.19: 3110100-3110100 on Build GRCh38
rs150293267 Chr.19: 3110114-3110114 on Build GRCh38
rs374969385 Chr.19: 3110121-3110121 on Build GRCh38
rs587777021 Chr.19: 3110190-3110190 on Build GRCh38
rs762714189 Chr.19: 3110152-3110152 on Build GRCh38
rs7255519 Chr.19: 3110125-3110125 on Build GRCh38
rs775915105 Chr.19: 3110139-3110139 on Build GRCh38
rs576887084 Chr.19: 3110207-3110207 on Build GRCh38
rs868681560 Chr.19: 3110209-3110209 on Build GRCh38
rs938891774 Chr.19: 3110086-3110086 on Build GRCh38
rs759067107 Chr.19: 3110162-3110162 on Build GRCh38
rs571571444 Chr.19: 3110127-3110127 on Build GRCh38
rs377224539 Chr.19: 3110163-3110163 on Build GRCh38
rs147570504 Chr.19: 3110156-3110156 on Build GRCh38
rs1056360254 Chr.19: 3110097-3110097 on Build GRCh38
rs149108190 Chr.19: 3110174-3110174 on Build GRCh38
rs563944908 Chr.19: 3110140-3110140 on Build GRCh38
rs779008056 Chr.19: 3110129-3110129 on Build GRCh38
rs776843648 Chr.19: 3110150-3110150 on Build GRCh38
rs781234448 Chr.19: 3110119-3110119 on Build GRCh38
rs761054897 Chr.19: 3110099-3110099 on Build GRCh38
rs769129067 Chr.19: 3110143-3110143 on Build GRCh38
rs587777707 Chr.19: 3110191-3110191 on Build GRCh38
rs755663461 Chr.19: 3110204-3110204 on Build GRCh38
rs200107023 Chr.19: 3110113-3110113 on Build GRCh38
rs368095546 Chr.19: 3110101-3110101 on Build GRCh38
rs752079029 Chr.19: 3110145-3110145 on Build GRCh38
rs964177853 Chr.19: 3110132-3110132 on Build GRCh38
rs933651595 Chr.19: 3110108-3110108 on Build GRCh38
rs745912473 Chr.19: 3110133-3110133 on Build GRCh38
rs777673683 Chr.19: 3110195-3110195 on Build GRCh38
rs61731116 Chr.19: 3110201-3110201 on Build GRCh38
rs766042469 Chr.19: 3110106-3110106 on Build GRCh38
rs147528229 Chr.19: 3110205-3110205 on Build GRCh38
rs755993488 Chr.19: 3110179-3110179 on Build GRCh38
rs371983140 Chr.19: 3110120-3110120 on Build GRCh38

View Full Product Details