Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46445259-46445404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
UniGene
Hs.529517
Gene Chromosome Location
Chr.3: 46436005-46485234 on Build GRCh38
Transcripts
NM_001199149.1 NM_001321121.1 NM_001321122.1 NM_002343.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6886525 c.1416G>A p.V472V Chr.3: 46445378-46445378 on Build GRCh38
6375433 c.1487T>A p.F496Y Chr.3: 46445307-46445307 on Build GRCh38
6341764 c.1454C>A p.T485N Chr.3: 46445340-46445340 on Build GRCh38
1214085 c.1441G>A p.A481T Chr.3: 46445353-46445353 on Build GRCh38
3594529 c.1451G>A p.R484K Chr.3: 46445343-46445343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764079632 Chr.3: 46445285-46445285 on Build GRCh38
rs1027361912 Chr.3: 46445333-46445333 on Build GRCh38
rs775399259 Chr.3: 46445322-46445322 on Build GRCh38
rs781138813 Chr.3: 46445295-46445295 on Build GRCh38
rs573022414 Chr.3: 46445351-46445351 on Build GRCh38
rs893113672 Chr.3: 46445281-46445281 on Build GRCh38
rs1020815714 Chr.3: 46445301-46445301 on Build GRCh38
rs751124361 Chr.3: 46445285-46445285 on Build GRCh38
rs773767772 Chr.3: 46445342-46445342 on Build GRCh38
rs777559903 Chr.3: 46445298-46445298 on Build GRCh38
rs878897245 Chr.3: 46445278-46445278 on Build GRCh38
rs777155554 Chr.3: 46445325-46445325 on Build GRCh38
rs762410872 Chr.3: 46445328-46445328 on Build GRCh38
rs368845863 Chr.3: 46445357-46445357 on Build GRCh38
rs200127456 Chr.3: 46445316-46445316 on Build GRCh38
rs762688257 Chr.3: 46445344-46445344 on Build GRCh38
rs879221135 Chr.3: 46445280-46445280 on Build GRCh38
rs867248457 Chr.3: 46445306-46445306 on Build GRCh38
rs754476078 Chr.3: 46445353-46445353 on Build GRCh38
rs1008461786 Chr.3: 46445282-46445282 on Build GRCh38
rs540544807 Chr.3: 46445354-46445354 on Build GRCh38
rs151143805 Chr.3: 46445297-46445297 on Build GRCh38
rs762558745 Chr.3: 46445288-46445288 on Build GRCh38
rs147094293 Chr.3: 46445350-46445350 on Build GRCh38
rs555018656 Chr.3: 46445364-46445364 on Build GRCh38
rs768640889 Chr.3: 46445324-46445324 on Build GRCh38
rs377248291 Chr.3: 46445319-46445319 on Build GRCh38
rs760857395 Chr.3: 46445323-46445323 on Build GRCh38
rs375348204 Chr.3: 46445327-46445327 on Build GRCh38
rs754853124 Chr.3: 46445294-46445294 on Build GRCh38
rs966137454 Chr.3: 46445346-46445346 on Build GRCh38
rs150320764 Chr.3: 46445302-46445302 on Build GRCh38
rs765635574 Chr.3: 46445334-46445334 on Build GRCh38
rs746078900 Chr.3: 46445312-46445312 on Build GRCh38

View Full Product Details