Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22109139-22109376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT18
Entrez Gene ID
79873
Gene Name
nudix hydrolase 18
Gene Aliases
MTH3
UniGene
Hs.527101
Gene Chromosome Location
Chr.8: 22106872-22110355 on Build GRCh38
Transcripts
NM_024815.3 XM_011544650.1

SNP Detail

SNP ID Chromosome Location
rs915538473 Chr.8: 22109231-22109231 on Build GRCh38
rs1041210406 Chr.8: 22109312-22109312 on Build GRCh38
rs200040808 Chr.8: 22109198-22109198 on Build GRCh38
rs768449580 Chr.8: 22109293-22109293 on Build GRCh38
rs367707934 Chr.8: 22109247-22109247 on Build GRCh38
rs758327673 Chr.8: 22109207-22109207 on Build GRCh38
rs753534345 Chr.8: 22109199-22109199 on Build GRCh38
rs536801022 Chr.8: 22109276-22109276 on Build GRCh38
rs781320705 Chr.8: 22109306-22109306 on Build GRCh38
rs957069904 Chr.8: 22109169-22109169 on Build GRCh38
rs796630966 Chr.8: 22109198-22109198 on Build GRCh38
rs894098030 Chr.8: 22109334-22109334 on Build GRCh38
rs1011316538 Chr.8: 22109359-22109359 on Build GRCh38
rs779995953 Chr.8: 22109210-22109210 on Build GRCh38
rs555204097 Chr.8: 22109295-22109295 on Build GRCh38
rs949739977 Chr.8: 22109248-22109248 on Build GRCh38
rs576023621 Chr.8: 22109243-22109243 on Build GRCh38
rs748396985 Chr.8: 22109313-22109313 on Build GRCh38
rs773317516 Chr.8: 22109360-22109360 on Build GRCh38
rs763783466 Chr.8: 22109187-22109187 on Build GRCh38
rs57316099 Chr.8: 22109190-22109190 on Build GRCh38
rs997068439 Chr.8: 22109323-22109323 on Build GRCh38
rs751986907 Chr.8: 22109305-22109305 on Build GRCh38
rs769967120 Chr.8: 22109342-22109342 on Build GRCh38
rs901354470 Chr.8: 22109316-22109316 on Build GRCh38
rs988964869 Chr.8: 22109213-22109213 on Build GRCh38
rs573370055 Chr.8: 22109352-22109352 on Build GRCh38
rs778475978 Chr.8: 22109203-22109203 on Build GRCh38
rs749953404 Chr.8: 22109205-22109205 on Build GRCh38
rs939775322 Chr.8: 22109310-22109310 on Build GRCh38
rs760002878 Chr.8: 22109177-22109177 on Build GRCh38
rs756931423 Chr.8: 22109200-22109200 on Build GRCh38
rs1048844929 Chr.8: 22109321-22109321 on Build GRCh38
rs927188324 Chr.8: 22109281-22109281 on Build GRCh38
rs371618895 Chr.8: 22109325-22109325 on Build GRCh38
rs938578280 Chr.8: 22109166-22109166 on Build GRCh38

View Full Product Details