Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131845300-131845549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP17
Entrez Gene ID
4326
Gene Name
matrix metallopeptidase 17
Gene Aliases
MMP-17, MT4-MMP, MT4MMP, MTMMP4
UniGene
Hs.709245
Gene Chromosome Location
Chr.12: 131828393-131851771 on Build GRCh38
Transcripts
NM_016155.5 XM_011538355.2 XM_011538356.2 XM_011538357.2 XM_017019307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895932 c.1071G>T p.L357L Chr.12: 131845316-131845316 on Build GRCh38
4797395 c.1065G>A p.K355K Chr.12: 131845310-131845310 on Build GRCh38
4702685 c.1158C>T p.D386D Chr.12: 131845403-131845403 on Build GRCh38
1605799 c.1153G>T p.V385L Chr.12: 131845398-131845398 on Build GRCh38
3359625 c.1171C>T p.R391C Chr.12: 131845416-131845416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79420934 Chr.12: 131845484-131845484 on Build GRCh38
rs902462730 Chr.12: 131845508-131845508 on Build GRCh38
rs556656006 Chr.12: 131845497-131845497 on Build GRCh38
rs570136987 Chr.12: 131845381-131845381 on Build GRCh38
rs775074248 Chr.12: 131845416-131845416 on Build GRCh38
rs745512111 Chr.12: 131845463-131845463 on Build GRCh38
rs760943298 Chr.12: 131845428-131845428 on Build GRCh38
rs769327440 Chr.12: 131845360-131845360 on Build GRCh38
rs1002281592 Chr.12: 131845520-131845520 on Build GRCh38
rs774317644 Chr.12: 131845419-131845419 on Build GRCh38
rs766447425 Chr.12: 131845431-131845431 on Build GRCh38
rs35490215 Chr.12: 131845461-131845462 on Build GRCh38
rs367543935 Chr.12: 131845332-131845332 on Build GRCh38
rs758748339 Chr.12: 131845441-131845441 on Build GRCh38
rs775628487 Chr.12: 131845362-131845362 on Build GRCh38
rs138124605 Chr.12: 131845389-131845389 on Build GRCh38
rs905368955 Chr.12: 131845513-131845513 on Build GRCh38
rs766552267 Chr.12: 131845487-131845487 on Build GRCh38
rs376752923 Chr.12: 131845466-131845466 on Build GRCh38
rs776238884 Chr.12: 131845350-131845350 on Build GRCh38
rs908144658 Chr.12: 131845456-131845456 on Build GRCh38
rs1006418770 Chr.12: 131845334-131845334 on Build GRCh38
rs745522975 Chr.12: 131845406-131845406 on Build GRCh38
rs1036653826 Chr.12: 131845356-131845356 on Build GRCh38
rs754540566 Chr.12: 131845320-131845320 on Build GRCh38
rs976585568 Chr.12: 131845442-131845442 on Build GRCh38
rs754404867 Chr.12: 131845385-131845385 on Build GRCh38
rs750420686 Chr.12: 131845331-131845331 on Build GRCh38
rs751898074 Chr.12: 131845326-131845326 on Build GRCh38
rs116392475 Chr.12: 131845465-131845465 on Build GRCh38
rs61753769 Chr.12: 131845377-131845377 on Build GRCh38
rs759011255 Chr.12: 131845353-131845353 on Build GRCh38
rs113706780 Chr.12: 131845397-131845397 on Build GRCh38
rs371736817 Chr.12: 131845436-131845436 on Build GRCh38
rs372018945 Chr.12: 131845413-131845413 on Build GRCh38
rs774477032 Chr.12: 131845471-131845471 on Build GRCh38
rs115545070 Chr.12: 131845498-131845498 on Build GRCh38
rs778678410 Chr.12: 131845321-131845321 on Build GRCh38
rs753228030 Chr.12: 131845437-131845437 on Build GRCh38
rs748100613 Chr.12: 131845476-131845476 on Build GRCh38
rs763984558 Chr.12: 131845371-131845371 on Build GRCh38
rs144769344 Chr.12: 131845363-131845363 on Build GRCh38
rs368806591 Chr.12: 131845468-131845468 on Build GRCh38
rs372008515 Chr.12: 131845530-131845530 on Build GRCh38
rs750666741 Chr.12: 131845425-131845425 on Build GRCh38
rs565985941 Chr.12: 131845404-131845404 on Build GRCh38
rs113204400 Chr.12: 131845403-131845403 on Build GRCh38
rs138777246 Chr.12: 131845373-131845373 on Build GRCh38
rs564370929 Chr.12: 131845319-131845319 on Build GRCh38
rs762997045 Chr.12: 131845417-131845417 on Build GRCh38
rs769063273 Chr.12: 131845348-131845348 on Build GRCh38
rs765048138 Chr.12: 131845318-131845318 on Build GRCh38
rs376826045 Chr.12: 131845494-131845494 on Build GRCh38
rs140855643 Chr.12: 131845349-131845349 on Build GRCh38
rs932053223 Chr.12: 131845475-131845475 on Build GRCh38
rs774792186 Chr.12: 131845321-131845321 on Build GRCh38
rs537025414 Chr.12: 131845485-131845485 on Build GRCh38
rs146309998 Chr.12: 131845382-131845382 on Build GRCh38
rs200341756 Chr.12: 131845424-131845424 on Build GRCh38
rs368085983 Chr.12: 131845457-131845457 on Build GRCh38
rs1046519810 Chr.12: 131845512-131845512 on Build GRCh38
rs746323549 Chr.12: 131845412-131845412 on Build GRCh38
rs780463604 Chr.12: 131845337-131845337 on Build GRCh38
rs781344733 Chr.12: 131845327-131845327 on Build GRCh38
rs768249546 Chr.12: 131845469-131845469 on Build GRCh38
rs767592241 Chr.12: 131845376-131845376 on Build GRCh38
rs371779544 Chr.12: 131845399-131845399 on Build GRCh38
rs763968267 Chr.12: 131845324-131845324 on Build GRCh38
rs12099616 Chr.12: 131845503-131845503 on Build GRCh38
rs374841041 Chr.12: 131845407-131845407 on Build GRCh38
rs759994504 Chr.12: 131845432-131845433 on Build GRCh38
rs753869953 Chr.12: 131845432-131845432 on Build GRCh38
rs749769535 Chr.12: 131845338-131845338 on Build GRCh38

View Full Product Details