Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28796493-28796742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS9
Entrez Gene ID
55756
Gene Name
integrator complex subunit 9
Gene Aliases
CPSF2L, INT9, RC74
UniGene
Hs.162397
Gene Chromosome Location
Chr.8: 28767658-28890226 on Build GRCh38
Transcripts
NM_001145159.2 NM_001172562.1 NM_018250.3 NR_026826.2 XM_006716356.2 XM_006716357.3 XM_011544573.1 XM_011544574.1 XM_011544575.2 XM_011544576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275575 c.788T>G p.L263R Chr.8: 28796612-28796612 on Build GRCh38
6113150 c.750G>A p.M250I Chr.8: 28796650-28796650 on Build GRCh38
6113151 c.765C>G p.L255L Chr.8: 28796635-28796635 on Build GRCh38
7416210 c.809C>A p.P270H Chr.8: 28796591-28796591 on Build GRCh38
3899452 c.774C>T p.S258S Chr.8: 28796626-28796626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557295921 Chr.8: 28796669-28796669 on Build GRCh38
rs777634823 Chr.8: 28796702-28796702 on Build GRCh38
rs572016374 Chr.8: 28796531-28796531 on Build GRCh38
rs990324377 Chr.8: 28796711-28796711 on Build GRCh38
rs143568689 Chr.8: 28796593-28796593 on Build GRCh38
rs951819819 Chr.8: 28796549-28796549 on Build GRCh38
rs747983258 Chr.8: 28796588-28796588 on Build GRCh38
rs762397337 Chr.8: 28796652-28796652 on Build GRCh38
rs749108353 Chr.8: 28796704-28796704 on Build GRCh38
rs78559720 Chr.8: 28796533-28796533 on Build GRCh38
rs375648052 Chr.8: 28796534-28796534 on Build GRCh38
rs368034981 Chr.8: 28796682-28796682 on Build GRCh38
rs781098699 Chr.8: 28796550-28796550 on Build GRCh38
rs764657235 Chr.8: 28796644-28796644 on Build GRCh38
rs376763337 Chr.8: 28796666-28796666 on Build GRCh38
rs761066475 Chr.8: 28796637-28796637 on Build GRCh38
rs775809172 Chr.8: 28796625-28796625 on Build GRCh38
rs777613993 Chr.8: 28796595-28796595 on Build GRCh38
rs941484725 Chr.8: 28796715-28796715 on Build GRCh38
rs752511667 Chr.8: 28796694-28796694 on Build GRCh38
rs748815707 Chr.8: 28796598-28796598 on Build GRCh38
rs368173552 Chr.8: 28796536-28796536 on Build GRCh38
rs924883748 Chr.8: 28796681-28796681 on Build GRCh38
rs535602450 Chr.8: 28796541-28796541 on Build GRCh38
rs867949633 Chr.8: 28796656-28796656 on Build GRCh38
rs372391267 Chr.8: 28796626-28796626 on Build GRCh38
rs771733417 Chr.8: 28796620-28796620 on Build GRCh38
rs74982893 Chr.8: 28796695-28796695 on Build GRCh38
rs914752578 Chr.8: 28796712-28796712 on Build GRCh38
rs773912263 Chr.8: 28796613-28796613 on Build GRCh38
rs758648883 Chr.8: 28796679-28796679 on Build GRCh38
rs975753137 Chr.8: 28796672-28796672 on Build GRCh38
rs770286628 Chr.8: 28796611-28796611 on Build GRCh38
rs777064393 Chr.8: 28796651-28796651 on Build GRCh38
rs369301059 Chr.8: 28796699-28796699 on Build GRCh38

View Full Product Details