Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3215505-3215778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1A1
Entrez Gene ID
8383
Gene Name
olfactory receptor family 1 subfamily A member 1
Gene Aliases
OR17-7
UniGene
Hs.532688
Gene Chromosome Location
Chr.17: 3215621-3216550 on Build GRCh38
Transcripts
NM_014565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516969 c.23C>A p.S8Y Chr.17: 3215643-3215643 on Build GRCh38
1382456 c.44T>A p.L15Q Chr.17: 3215664-3215664 on Build GRCh38
4556679 c.69G>A p.Q23Q Chr.17: 3215689-3215689 on Build GRCh38
6340915 c.96G>T p.L32F Chr.17: 3215716-3215716 on Build GRCh38
4065435 c.110T>C p.I37T Chr.17: 3215730-3215730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029042752 Chr.17: 3215684-3215684 on Build GRCh38
rs777953742 Chr.17: 3215609-3215609 on Build GRCh38
rs372506808 Chr.17: 3215756-3215756 on Build GRCh38
rs4325604 Chr.17: 3215714-3215714 on Build GRCh38
rs957670070 Chr.17: 3215720-3215720 on Build GRCh38
rs773114544 Chr.17: 3215734-3215734 on Build GRCh38
rs774730866 Chr.17: 3215746-3215746 on Build GRCh38
rs916757902 Chr.17: 3215548-3215548 on Build GRCh38
rs763280963 Chr.17: 3215659-3215659 on Build GRCh38
rs376198426 Chr.17: 3215727-3215727 on Build GRCh38
rs765110007 Chr.17: 3215646-3215646 on Build GRCh38
rs766207040 Chr.17: 3215742-3215742 on Build GRCh38
rs758793744 Chr.17: 3215552-3215552 on Build GRCh38
rs373650324 Chr.17: 3215716-3215716 on Build GRCh38
rs768175847 Chr.17: 3215721-3215721 on Build GRCh38
rs776562506 Chr.17: 3215636-3215636 on Build GRCh38
rs767650485 Chr.17: 3215672-3215672 on Build GRCh38
rs916331363 Chr.17: 3215735-3215735 on Build GRCh38
rs376685479 Chr.17: 3215595-3215595 on Build GRCh38
rs948182783 Chr.17: 3215554-3215554 on Build GRCh38
rs1004914986 Chr.17: 3215618-3215618 on Build GRCh38
rs756541505 Chr.17: 3215692-3215692 on Build GRCh38
rs745568148 Chr.17: 3215617-3215617 on Build GRCh38
rs978009550 Chr.17: 3215718-3215718 on Build GRCh38
rs748602940 Chr.17: 3215582-3215582 on Build GRCh38
rs775031743 Chr.17: 3215622-3215622 on Build GRCh38
rs761605171 Chr.17: 3215637-3215637 on Build GRCh38
rs778242671 Chr.17: 3215708-3215708 on Build GRCh38
rs571378546 Chr.17: 3215730-3215730 on Build GRCh38
rs762647120 Chr.17: 3215739-3215739 on Build GRCh38
rs1011785074 Chr.17: 3215677-3215677 on Build GRCh38
rs369213483 Chr.17: 3215621-3215621 on Build GRCh38
rs1030638500 Chr.17: 3215719-3215719 on Build GRCh38
rs1046684493 Chr.17: 3215562-3215562 on Build GRCh38
rs201744204 Chr.17: 3215669-3215669 on Build GRCh38
rs751751647 Chr.17: 3215663-3215663 on Build GRCh38
rs756518635 Chr.17: 3215589-3215589 on Build GRCh38
rs772848853 Chr.17: 3215657-3215657 on Build GRCh38
rs1007125749 Chr.17: 3215704-3215704 on Build GRCh38
rs779677197 Chr.17: 3215715-3215715 on Build GRCh38
rs370727950 Chr.17: 3215633-3215633 on Build GRCh38
rs759786709 Chr.17: 3215751-3215751 on Build GRCh38
rs1053379277 Chr.17: 3215648-3215648 on Build GRCh38
rs952611430 Chr.17: 3215549-3215549 on Build GRCh38
rs919722437 Chr.17: 3215545-3215545 on Build GRCh38
rs781762484 Chr.17: 3215578-3215578 on Build GRCh38
rs749594794 Chr.17: 3215713-3215713 on Build GRCh38
rs768994377 Chr.17: 3215725-3215725 on Build GRCh38
rs1036754516 Chr.17: 3215656-3215656 on Build GRCh38
rs994929821 Chr.17: 3215675-3215675 on Build GRCh38
rs4374201 Chr.17: 3215755-3215755 on Build GRCh38
rs753272824 Chr.17: 3215691-3215691 on Build GRCh38
rs768648171 Chr.17: 3215630-3215630 on Build GRCh38
rs776078526 Chr.17: 3215725-3215725 on Build GRCh38
rs982676054 Chr.17: 3215750-3215750 on Build GRCh38

View Full Product Details