Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55469306-55469551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6C70
Entrez Gene ID
390327
Gene Name
olfactory receptor family 6 subfamily C member 70
Gene Aliases
-
UniGene
Hs.632718
Gene Chromosome Location
Chr.12: 55469200-55470138 on Build GRCh38
Transcripts
NM_001005499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5805166 c.629T>A p.F210Y Chr.12: 55469510-55469510 on Build GRCh38
3463096 c.695A>T p.K232M Chr.12: 55469444-55469444 on Build GRCh38
4567504 c.740_741CC>TT p.S247F Chr.12: 55469398-55469399 on Build GRCh38
5721976 c.672G>A p.L224L Chr.12: 55469467-55469467 on Build GRCh38
3463094 c.811G>A p.G271R Chr.12: 55469328-55469328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754693677 Chr.12: 55469403-55469403 on Build GRCh38
rs562989194 Chr.12: 55469426-55469426 on Build GRCh38
rs776105671 Chr.12: 55469353-55469353 on Build GRCh38
rs756217780 Chr.12: 55469506-55469506 on Build GRCh38
rs141797471 Chr.12: 55469504-55469504 on Build GRCh38
rs772723628 Chr.12: 55469339-55469339 on Build GRCh38
rs760236906 Chr.12: 55469350-55469350 on Build GRCh38
rs770957229 Chr.12: 55469502-55469502 on Build GRCh38
rs566507422 Chr.12: 55469499-55469499 on Build GRCh38
rs747700479 Chr.12: 55469435-55469435 on Build GRCh38
rs961433376 Chr.12: 55469401-55469401 on Build GRCh38
rs774762314 Chr.12: 55469485-55469485 on Build GRCh38
rs1008053242 Chr.12: 55469386-55469386 on Build GRCh38
rs60683621 Chr.12: 55469440-55469440 on Build GRCh38
rs74893107 Chr.12: 55469398-55469398 on Build GRCh38
rs751665007 Chr.12: 55469361-55469361 on Build GRCh38
rs149987906 Chr.12: 55469411-55469411 on Build GRCh38
rs753654132 Chr.12: 55469508-55469508 on Build GRCh38
rs770478050 Chr.12: 55469452-55469452 on Build GRCh38
rs764549180 Chr.12: 55469360-55469360 on Build GRCh38
rs767906498 Chr.12: 55469378-55469378 on Build GRCh38
rs149166293 Chr.12: 55469460-55469460 on Build GRCh38
rs140433772 Chr.12: 55469354-55469354 on Build GRCh38
rs756299390 Chr.12: 55469384-55469384 on Build GRCh38
rs879086585 Chr.12: 55469418-55469418 on Build GRCh38
rs763520707 Chr.12: 55469479-55469479 on Build GRCh38
rs377195328 Chr.12: 55469367-55469367 on Build GRCh38
rs529572052 Chr.12: 55469389-55469389 on Build GRCh38
rs769426525 Chr.12: 55469475-55469475 on Build GRCh38
rs766514494 Chr.12: 55469507-55469507 on Build GRCh38
rs978207655 Chr.12: 55469515-55469515 on Build GRCh38
rs370700627 Chr.12: 55469491-55469491 on Build GRCh38
rs771374972 Chr.12: 55469450-55469450 on Build GRCh38
rs890986204 Chr.12: 55469376-55469376 on Build GRCh38
rs764306375 Chr.12: 55469481-55469481 on Build GRCh38
rs779829960 Chr.12: 55469391-55469391 on Build GRCh38
rs778775806 Chr.12: 55469522-55469522 on Build GRCh38
rs750524022 Chr.12: 55469379-55469379 on Build GRCh38
rs771702337 Chr.12: 55469436-55469436 on Build GRCh38
rs749383569 Chr.12: 55469486-55469487 on Build GRCh38
rs983481956 Chr.12: 55469390-55469390 on Build GRCh38
rs754959565 Chr.12: 55469514-55469514 on Build GRCh38

View Full Product Details