Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17420641-17420902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC2
Entrez Gene ID
55920
Gene Name
regulator of chromosome condensation 2
Gene Aliases
TD-60
UniGene
Hs.380857
Gene Chromosome Location
Chr.1: 17406755-17439754 on Build GRCh38
Transcripts
NM_001136204.2 NM_018715.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1287571 c.779C>G p.A260G Chr.1: 17420794-17420794 on Build GRCh38
6540138 c.781T>A p.C261S Chr.1: 17420792-17420792 on Build GRCh38
463482 c.859G>C p.G287R Chr.1: 17420714-17420714 on Build GRCh38
5907161 c.818G>A p.G273E Chr.1: 17420755-17420755 on Build GRCh38
5052826 c.782G>A p.C261Y Chr.1: 17420791-17420791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022975662 Chr.1: 17420816-17420816 on Build GRCh38
rs771202293 Chr.1: 17420694-17420695 on Build GRCh38
rs779976827 Chr.1: 17420778-17420778 on Build GRCh38
rs200812013 Chr.1: 17420855-17420863 on Build GRCh38
rs376910701 Chr.1: 17420802-17420802 on Build GRCh38
rs763648052 Chr.1: 17420690-17420690 on Build GRCh38
rs781493205 Chr.1: 17420803-17420803 on Build GRCh38
rs764994560 Chr.1: 17420740-17420740 on Build GRCh38
rs749596397 Chr.1: 17420828-17420828 on Build GRCh38
rs778165911 Chr.1: 17420817-17420817 on Build GRCh38
rs776166818 Chr.1: 17420860-17420860 on Build GRCh38
rs372464208 Chr.1: 17420838-17420838 on Build GRCh38
rs768122567 Chr.1: 17420849-17420849 on Build GRCh38
rs751466244 Chr.1: 17420787-17420787 on Build GRCh38
rs201195434 Chr.1: 17420863-17420863 on Build GRCh38
rs755073915 Chr.1: 17420798-17420798 on Build GRCh38
rs770193380 Chr.1: 17420809-17420809 on Build GRCh38
rs750168150 Chr.1: 17420746-17420746 on Build GRCh38
rs748353564 Chr.1: 17420807-17420807 on Build GRCh38
rs864622010 Chr.1: 17420791-17420791 on Build GRCh38
rs763349645 Chr.1: 17420664-17420664 on Build GRCh38
rs774903662 Chr.1: 17420844-17420844 on Build GRCh38
rs776996001 Chr.1: 17420832-17420832 on Build GRCh38
rs764828110 Chr.1: 17420867-17420867 on Build GRCh38
rs761491892 Chr.1: 17420702-17420702 on Build GRCh38
rs1052510049 Chr.1: 17420674-17420674 on Build GRCh38
rs1013380471 Chr.1: 17420748-17420748 on Build GRCh38
rs775589985 Chr.1: 17420855-17420856 on Build GRCh38
rs747472776 Chr.1: 17420688-17420688 on Build GRCh38
rs540184917 Chr.1: 17420747-17420747 on Build GRCh38
rs753480467 Chr.1: 17420696-17420696 on Build GRCh38
rs1016517807 Chr.1: 17420845-17420845 on Build GRCh38
rs760171739 Chr.1: 17420685-17420685 on Build GRCh38
rs771423422 Chr.1: 17420666-17420666 on Build GRCh38
rs760025449 Chr.1: 17420848-17420848 on Build GRCh38
rs533684481 Chr.1: 17420851-17420852 on Build GRCh38
rs184740592 Chr.1: 17420683-17420683 on Build GRCh38

View Full Product Details