Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46040318-46040561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A5
Entrez Gene ID
57468
Gene Name
solute carrier family 12 member 5
Gene Aliases
EIEE34, EIG14, KCC2, hKCC2
UniGene
Hs.21413
Gene Chromosome Location
Chr.20: 46021690-46060150 on Build GRCh38
Transcripts
NM_001134771.1 NM_020708.4 XM_017027981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1225880 c.725C>T p.A242V Chr.20: 46040485-46040485 on Build GRCh38
5914327 c.613-6C>T p.? Chr.20: 46040367-46040367 on Build GRCh38
6207894 c.724G>A p.A242T Chr.20: 46040415-46040415 on Build GRCh38
6858321 c.625C>T p.P209S Chr.20: 46040385-46040385 on Build GRCh38
6579130 c.690C>G p.L230L Chr.20: 46040381-46040381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915523015 Chr.20: 46040532-46040532 on Build GRCh38
rs533415684 Chr.20: 46040435-46040435 on Build GRCh38
rs372753758 Chr.20: 46040507-46040507 on Build GRCh38
rs370495847 Chr.20: 46040397-46040397 on Build GRCh38
rs747994334 Chr.20: 46040342-46040342 on Build GRCh38
rs777601251 Chr.20: 46040510-46040510 on Build GRCh38
rs771779385 Chr.20: 46040359-46040359 on Build GRCh38
rs745445077 Chr.20: 46040356-46040356 on Build GRCh38
rs749289545 Chr.20: 46040345-46040345 on Build GRCh38
rs777269119 Chr.20: 46040458-46040458 on Build GRCh38
rs780806926 Chr.20: 46040339-46040339 on Build GRCh38
rs913680770 Chr.20: 46040390-46040390 on Build GRCh38
rs773863359 Chr.20: 46040351-46040351 on Build GRCh38
rs765343735 Chr.20: 46040460-46040460 on Build GRCh38
rs201198034 Chr.20: 46040494-46040494 on Build GRCh38
rs965878928 Chr.20: 46040355-46040355 on Build GRCh38
rs764620289 Chr.20: 46040431-46040431 on Build GRCh38
rs1046062288 Chr.20: 46040522-46040522 on Build GRCh38
rs575958914 Chr.20: 46040343-46040343 on Build GRCh38
rs766598411 Chr.20: 46040483-46040483 on Build GRCh38
rs140635575 Chr.20: 46040388-46040388 on Build GRCh38
rs201729629 Chr.20: 46040511-46040511 on Build GRCh38
rs750550342 Chr.20: 46040471-46040471 on Build GRCh38
rs763034305 Chr.20: 46040476-46040476 on Build GRCh38
rs775057000 Chr.20: 46040367-46040367 on Build GRCh38
rs200570265 Chr.20: 46040459-46040459 on Build GRCh38
rs932821826 Chr.20: 46040429-46040429 on Build GRCh38

View Full Product Details