Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89105408-89105649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PM20D2
Entrez Gene ID
135293
Gene Name
peptidase M20 domain containing 2
Gene Aliases
ACY1L2, bA63L7.3
UniGene
Hs.356247
Gene Chromosome Location
Chr.6: 89096879-89165569 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Gene Symbol
SRSF12
Entrez Gene ID
135295
Gene Name
serine and arginine rich splicing factor 12
Gene Aliases
SFRS13B, SFRS19, SRrp35
UniGene
Hs.254414
Gene Chromosome Location
Chr.6: 89095957-89118081 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5688121 c.198T>G p.D66E Chr.6: 89105503-89105503 on Build GRCh38
6224183 c.269G>A p.R90H Chr.6: 89105432-89105432 on Build GRCh38
7269534 c.200C>A p.A67D Chr.6: 89105501-89105501 on Build GRCh38
3178023 c.184C>T p.R62* Chr.6: 89105517-89105517 on Build GRCh38
236318 c.219delA p.K74fs*23 Chr.6: 89105482-89105482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768267669 Chr.6: 89105517-89105517 on Build GRCh38
rs988948512 Chr.6: 89105558-89105558 on Build GRCh38
rs766782791 Chr.6: 89105461-89105461 on Build GRCh38
rs766238628 Chr.6: 89105469-89105469 on Build GRCh38
rs772601317 Chr.6: 89105577-89105577 on Build GRCh38
rs755198433 Chr.6: 89105465-89105465 on Build GRCh38
rs369780296 Chr.6: 89105564-89105564 on Build GRCh38
rs763514078 Chr.6: 89105578-89105578 on Build GRCh38
rs560927391 Chr.6: 89105490-89105490 on Build GRCh38
rs933374923 Chr.6: 89105570-89105570 on Build GRCh38
rs980242192 Chr.6: 89105588-89105588 on Build GRCh38
rs536856733 Chr.6: 89105452-89105452 on Build GRCh38
rs769288615 Chr.6: 89105565-89105565 on Build GRCh38
rs189132529 Chr.6: 89105609-89105609 on Build GRCh38
rs750312037 Chr.6: 89105500-89105500 on Build GRCh38
rs886938009 Chr.6: 89105592-89105592 on Build GRCh38
rs373177602 Chr.6: 89105573-89105573 on Build GRCh38
rs756277592 Chr.6: 89105489-89105489 on Build GRCh38
rs375463727 Chr.6: 89105516-89105516 on Build GRCh38
rs752000227 Chr.6: 89105462-89105462 on Build GRCh38
rs752685727 Chr.6: 89105486-89105486 on Build GRCh38
rs373163602 Chr.6: 89105509-89105509 on Build GRCh38
rs979661980 Chr.6: 89105432-89105432 on Build GRCh38
rs17856643 Chr.6: 89105513-89105513 on Build GRCh38
rs758367069 Chr.6: 89105502-89105502 on Build GRCh38
rs1033104206 Chr.6: 89105466-89105466 on Build GRCh38
rs780966903 Chr.6: 89105541-89105541 on Build GRCh38
rs747166270 Chr.6: 89105475-89105475 on Build GRCh38
rs925587111 Chr.6: 89105440-89105440 on Build GRCh38
rs866909358 Chr.6: 89105493-89105493 on Build GRCh38
rs917035584 Chr.6: 89105568-89105568 on Build GRCh38
rs1004023760 Chr.6: 89105616-89105616 on Build GRCh38

View Full Product Details