Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90954586-90954811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCER1
Entrez Gene ID
196477
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12
UniGene
Hs.128574
Gene Chromosome Location
Chr.12: 90952215-90955176 on Build GRCh38
Transcripts
NM_152638.3 NR_130711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4940819 c.78G>A p.W26* Chr.12: 90954665-90954665 on Build GRCh38
4919993 c.117T>C p.H39H Chr.12: 90954626-90954626 on Build GRCh38
4045344 c.54C>T p.G18G Chr.12: 90954689-90954689 on Build GRCh38
4045343 c.141G>A p.A47A Chr.12: 90954602-90954602 on Build GRCh38
6622740 c.119G>A p.R40Q Chr.12: 90954624-90954624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953850152 Chr.12: 90954772-90954772 on Build GRCh38
rs1028196132 Chr.12: 90954747-90954747 on Build GRCh38
rs778176162 Chr.12: 90954757-90954757 on Build GRCh38
rs117919138 Chr.12: 90954779-90954779 on Build GRCh38
rs772988715 Chr.12: 90954697-90954697 on Build GRCh38
rs145909006 Chr.12: 90954656-90954656 on Build GRCh38
rs911809425 Chr.12: 90954682-90954682 on Build GRCh38
rs757526649 Chr.12: 90954778-90954778 on Build GRCh38
rs376363819 Chr.12: 90954689-90954689 on Build GRCh38
rs754030690 Chr.12: 90954760-90954760 on Build GRCh38
rs768072610 Chr.12: 90954732-90954732 on Build GRCh38
rs774455950 Chr.12: 90954655-90954655 on Build GRCh38
rs750382205 Chr.12: 90954686-90954687 on Build GRCh38
rs561661034 Chr.12: 90954743-90954743 on Build GRCh38
rs902954665 Chr.12: 90954787-90954788 on Build GRCh38
rs756222901 Chr.12: 90954648-90954648 on Build GRCh38
rs761676354 Chr.12: 90954692-90954692 on Build GRCh38
rs370822454 Chr.12: 90954635-90954635 on Build GRCh38
rs753463782 Chr.12: 90954680-90954682 on Build GRCh38
rs762733226 Chr.12: 90954707-90954707 on Build GRCh38
rs764977047 Chr.12: 90954607-90954607 on Build GRCh38
rs752078222 Chr.12: 90954630-90954630 on Build GRCh38
rs770995521 Chr.12: 90954654-90954654 on Build GRCh38
rs867467733 Chr.12: 90954625-90954625 on Build GRCh38
rs781358100 Chr.12: 90954642-90954642 on Build GRCh38
rs778479718 Chr.12: 90954681-90954692 on Build GRCh38
rs748576772 Chr.12: 90954647-90954647 on Build GRCh38
rs986088543 Chr.12: 90954662-90954662 on Build GRCh38
rs751133712 Chr.12: 90954713-90954713 on Build GRCh38
rs756753753 Chr.12: 90954679-90954680 on Build GRCh38
rs540142735 Chr.12: 90954683-90954683 on Build GRCh38
rs189723536 Chr.12: 90954694-90954694 on Build GRCh38
rs557158788 Chr.12: 90954649-90954649 on Build GRCh38
rs541896912 Chr.12: 90954710-90954710 on Build GRCh38
rs748954628 Chr.12: 90954728-90954728 on Build GRCh38
rs755395979 Chr.12: 90954715-90954715 on Build GRCh38
rs749568876 Chr.12: 90954650-90954650 on Build GRCh38
rs752945465 Chr.12: 90954733-90954733 on Build GRCh38
rs1003809395 Chr.12: 90954628-90954628 on Build GRCh38
rs537131134 Chr.12: 90954615-90954615 on Build GRCh38
rs766046971 Chr.12: 90954621-90954621 on Build GRCh38

View Full Product Details