Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56143726-56143999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD55
Entrez Gene ID
79722
Gene Name
ankyrin repeat domain 55
Gene Aliases
-
UniGene
Hs.436214
Gene Chromosome Location
Chr.5: 56099680-56233359 on Build GRCh38
Transcripts
NM_024669.2 XM_011543646.1 XM_017009852.1 XM_017009853.1 XM_017009854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457199 c.507G>A p.A169A Chr.5: 56143906-56143906 on Build GRCh38
1328751 c.520C>G p.Q174E Chr.5: 56143893-56143893 on Build GRCh38
5927362 c.553G>A p.G185R Chr.5: 56143860-56143860 on Build GRCh38
6104512 c.579A>T p.K193N Chr.5: 56143834-56143834 on Build GRCh38
6600853 c.544C>T p.L182L Chr.5: 56143869-56143869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780373470 Chr.5: 56143952-56143952 on Build GRCh38
rs201143186 Chr.5: 56143801-56143801 on Build GRCh38
rs748664603 Chr.5: 56143908-56143908 on Build GRCh38
rs761204172 Chr.5: 56143928-56143928 on Build GRCh38
rs746434017 Chr.5: 56143841-56143841 on Build GRCh38
rs760126113 Chr.5: 56143790-56143790 on Build GRCh38
rs761590515 Chr.5: 56143755-56143755 on Build GRCh38
rs770568200 Chr.5: 56143973-56143973 on Build GRCh38
rs752432307 Chr.5: 56143812-56143812 on Build GRCh38
rs749900940 Chr.5: 56143780-56143787 on Build GRCh38
rs878964772 Chr.5: 56143859-56143859 on Build GRCh38
rs766339156 Chr.5: 56143792-56143792 on Build GRCh38
rs756492807 Chr.5: 56143941-56143944 on Build GRCh38
rs771590485 Chr.5: 56143765-56143765 on Build GRCh38
rs200820464 Chr.5: 56143956-56143956 on Build GRCh38
rs770077288 Chr.5: 56143852-56143852 on Build GRCh38
rs777497930 Chr.5: 56143961-56143961 on Build GRCh38
rs781588786 Chr.5: 56143857-56143857 on Build GRCh38
rs751710930 Chr.5: 56143821-56143821 on Build GRCh38
rs757537830 Chr.5: 56143907-56143907 on Build GRCh38
rs992779309 Chr.5: 56143819-56143819 on Build GRCh38
rs143369684 Chr.5: 56143806-56143806 on Build GRCh38
rs770990470 Chr.5: 56143890-56143890 on Build GRCh38
rs780505029 Chr.5: 56143845-56143845 on Build GRCh38
rs528754382 Chr.5: 56143900-56143900 on Build GRCh38
rs776109275 Chr.5: 56143882-56143882 on Build GRCh38
rs376260776 Chr.5: 56143813-56143813 on Build GRCh38
rs1042118636 Chr.5: 56143896-56143896 on Build GRCh38
rs749590597 Chr.5: 56143847-56143847 on Build GRCh38
rs199518538 Chr.5: 56143771-56143771 on Build GRCh38
rs770373905 Chr.5: 56143879-56143879 on Build GRCh38
rs760187463 Chr.5: 56143873-56143873 on Build GRCh38
rs767770861 Chr.5: 56143920-56143920 on Build GRCh38
rs764381439 Chr.5: 56143897-56143897 on Build GRCh38
rs777836712 Chr.5: 56143820-56143820 on Build GRCh38
rs772695526 Chr.5: 56143861-56143861 on Build GRCh38
rs777574747 Chr.5: 56143863-56143863 on Build GRCh38
rs755571114 Chr.5: 56143808-56143809 on Build GRCh38
rs746725233 Chr.5: 56143969-56143969 on Build GRCh38
rs150093349 Chr.5: 56143894-56143894 on Build GRCh38
rs202182721 Chr.5: 56143891-56143891 on Build GRCh38
rs756690823 Chr.5: 56143949-56143949 on Build GRCh38
rs762608186 Chr.5: 56143895-56143895 on Build GRCh38
rs200572856 Chr.5: 56143831-56143831 on Build GRCh38
rs371801644 Chr.5: 56143843-56143843 on Build GRCh38
rs368520889 Chr.5: 56143822-56143822 on Build GRCh38
rs750497480 Chr.5: 56143928-56143928 on Build GRCh38
rs148379117 Chr.5: 56143807-56143807 on Build GRCh38

View Full Product Details