Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42784016-42784263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNK4
Entrez Gene ID
65266
Gene Name
WNK lysine deficient protein kinase 4
Gene Aliases
PHA2B, PRKWNK4
UniGene
Hs.105448
Gene Chromosome Location
Chr.17: 42779701-42797066 on Build GRCh38
Transcripts
NM_001321299.1 NM_032387.4 XM_011525135.2 XM_017024962.1 XM_017024963.1 XM_017024964.1 XM_017024965.1 XM_017024966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579720 c.958G>A p.G320R Chr.17: 42784103-42784103 on Build GRCh38
6839407 c.947C>G p.S316C Chr.17: 42784092-42784092 on Build GRCh38
5481379 c.933G>A p.T311T Chr.17: 42784078-42784078 on Build GRCh38
7397907 c.890C>T p.P297L Chr.17: 42784035-42784035 on Build GRCh38
4066644 c.939T>C p.P313P Chr.17: 42784084-42784084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779031905 Chr.17: 42784069-42784069 on Build GRCh38
rs142280606 Chr.17: 42784130-42784130 on Build GRCh38
rs767158498 Chr.17: 42784108-42784108 on Build GRCh38
rs984286963 Chr.17: 42784123-42784123 on Build GRCh38
rs387907559 Chr.17: 42784118-42784118 on Build GRCh38
rs747424461 Chr.17: 42784049-42784049 on Build GRCh38
rs373585658 Chr.17: 42784157-42784157 on Build GRCh38
rs778048173 Chr.17: 42784046-42784046 on Build GRCh38
rs921379364 Chr.17: 42784045-42784045 on Build GRCh38
rs951793928 Chr.17: 42784161-42784161 on Build GRCh38
rs755245471 Chr.17: 42784122-42784122 on Build GRCh38
rs557180003 Chr.17: 42784169-42784169 on Build GRCh38
rs761672066 Chr.17: 42784103-42784103 on Build GRCh38
rs778377540 Chr.17: 42784147-42784147 on Build GRCh38
rs776352888 Chr.17: 42784063-42784063 on Build GRCh38
rs368501387 Chr.17: 42784086-42784086 on Build GRCh38
rs940837723 Chr.17: 42784238-42784238 on Build GRCh38
rs752803508 Chr.17: 42784135-42784135 on Build GRCh38
rs200256086 Chr.17: 42784078-42784078 on Build GRCh38
rs755199788 Chr.17: 42784060-42784060 on Build GRCh38
rs540080053 Chr.17: 42784214-42784214 on Build GRCh38
rs764038708 Chr.17: 42784084-42784084 on Build GRCh38
rs150813956 Chr.17: 42784205-42784205 on Build GRCh38
rs61754348 Chr.17: 42784090-42784090 on Build GRCh38
rs747410788 Chr.17: 42784163-42784163 on Build GRCh38
rs759883056 Chr.17: 42784103-42784103 on Build GRCh38
rs908064895 Chr.17: 42784225-42784225 on Build GRCh38
rs772677276 Chr.17: 42784037-42784037 on Build GRCh38
rs748824659 Chr.17: 42784195-42784195 on Build GRCh38
rs761729617 Chr.17: 42784195-42784195 on Build GRCh38
rs769410713 Chr.17: 42784176-42784176 on Build GRCh38
rs754147577 Chr.17: 42784109-42784109 on Build GRCh38
rs775070496 Chr.17: 42784190-42784190 on Build GRCh38
rs139202412 Chr.17: 42784082-42784082 on Build GRCh38
rs981757986 Chr.17: 42784240-42784240 on Build GRCh38
rs758505108 Chr.17: 42784142-42784142 on Build GRCh38
rs781400955 Chr.17: 42784172-42784172 on Build GRCh38
rs1036678369 Chr.17: 42784194-42784194 on Build GRCh38
rs759056532 Chr.17: 42784065-42784065 on Build GRCh38

View Full Product Details