Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 171063788-171064061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNIK
Entrez Gene ID
23043
Gene Name
TRAF2 and NCK interacting kinase
Gene Aliases
-
UniGene
Hs.34024
Gene Chromosome Location
Chr.3: 171058414-171460408 on Build GRCh38
Transcripts
NM_001161560.2 NM_001161561.2 NM_001161562.2 NM_001161563.2 NM_001161564.2 NM_001161565.2 NM_001161566.2 NM_015028.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
285847 c.4018C>T p.R1340* Chr.3: 171063946-171063946 on Build GRCh38
3427324 c.4006T>G p.F1336V Chr.3: 171063958-171063958 on Build GRCh38
402394 c.4071G>T p.M1357I Chr.3: 171063893-171063893 on Build GRCh38
4546568 c.4028G>A p.G1343E Chr.3: 171063936-171063936 on Build GRCh38
1566593 c.4008delT p.F1336fs*15 Chr.3: 171063956-171063956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760900860 Chr.3: 171063970-171063971 on Build GRCh38
rs757996859 Chr.3: 171063949-171063949 on Build GRCh38
rs199605404 Chr.3: 171063950-171063950 on Build GRCh38
rs371364550 Chr.3: 171063828-171063828 on Build GRCh38
rs1048322235 Chr.3: 171064005-171064005 on Build GRCh38
rs764869738 Chr.3: 171063985-171063985 on Build GRCh38
rs3821746 Chr.3: 171063823-171063823 on Build GRCh38
rs754260207 Chr.3: 171063864-171063864 on Build GRCh38
rs899324696 Chr.3: 171064018-171064018 on Build GRCh38
rs185162556 Chr.3: 171063850-171063850 on Build GRCh38
rs189969139 Chr.3: 171063946-171063946 on Build GRCh38
rs557723970 Chr.3: 171064026-171064026 on Build GRCh38
rs747342787 Chr.3: 171063841-171063841 on Build GRCh38
rs372474615 Chr.3: 171063959-171063959 on Build GRCh38
rs758943336 Chr.3: 171063835-171063835 on Build GRCh38
rs773329490 Chr.3: 171063887-171063887 on Build GRCh38
rs769168794 Chr.3: 171063849-171063849 on Build GRCh38
rs749947110 Chr.3: 171063832-171063832 on Build GRCh38
rs1037912118 Chr.3: 171064016-171064016 on Build GRCh38
rs770189582 Chr.3: 171063877-171063877 on Build GRCh38
rs748351400 Chr.3: 171063854-171063854 on Build GRCh38
rs763008088 Chr.3: 171063895-171063895 on Build GRCh38
rs771977662 Chr.3: 171063906-171063906 on Build GRCh38
rs547618278 Chr.3: 171063813-171063813 on Build GRCh38
rs749886771 Chr.3: 171064012-171064012 on Build GRCh38
rs375606742 Chr.3: 171063853-171063853 on Build GRCh38
rs761429782 Chr.3: 171063962-171063962 on Build GRCh38
rs780433221 Chr.3: 171063840-171063840 on Build GRCh38

View Full Product Details