Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155056806-155057045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
UniGene
Hs.312098
Gene Chromosome Location
Chr.1: 155051272-155062776 on Build GRCh38
Transcripts
NM_001261464.1 NM_001261465.1 NM_001261466.1 NM_003815.4 NM_207191.2 NM_207194.2 NM_207195.2 NM_207196.2 NM_207197.2 NR_048577.1 NR_048578.1 NR_048579.1 NR_040773.1

Gene Symbol
LOC100505666
Entrez Gene ID
100505666
Gene Name
uncharacterized LOC100505666
Gene Aliases
-
UniGene
Hs.657533
Gene Chromosome Location
Chr.1: 155045191-155063991 on Build GRCh38
Transcripts
NM_001261464.1 NM_001261465.1 NM_001261466.1 NM_003815.4 NM_207191.2 NM_207194.2 NM_207195.2 NM_207196.2 NM_207197.2 NR_048577.1 NR_048578.1 NR_048579.1 NR_040773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7003971 c.1032C>A p.S344S Chr.1: 155056985-155056985 on Build GRCh38
896758 c.1010C>T p.T337I Chr.1: 155056963-155056963 on Build GRCh38
3385226 c.1015A>G p.I339V Chr.1: 155056968-155056968 on Build GRCh38
7428565 c.1038A>T p.I346I Chr.1: 155056991-155056991 on Build GRCh38
4022929 c.1044T>C p.H348H Chr.1: 155056997-155056997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755597483 Chr.1: 155056993-155056993 on Build GRCh38
rs778708750 Chr.1: 155057020-155057020 on Build GRCh38
rs770054372 Chr.1: 155056954-155056954 on Build GRCh38
rs768018320 Chr.1: 155056989-155056989 on Build GRCh38
rs773464499 Chr.1: 155056956-155056956 on Build GRCh38
rs980459347 Chr.1: 155056864-155056864 on Build GRCh38
rs753489498 Chr.1: 155057011-155057011 on Build GRCh38
rs766729173 Chr.1: 155056975-155056975 on Build GRCh38
rs745658433 Chr.1: 155057021-155057021 on Build GRCh38
rs761977624 Chr.1: 155056950-155056950 on Build GRCh38
rs970650021 Chr.1: 155056838-155056838 on Build GRCh38
rs114509629 Chr.1: 155056863-155056863 on Build GRCh38
rs753271272 Chr.1: 155056992-155056992 on Build GRCh38
rs751892671 Chr.1: 155056979-155056979 on Build GRCh38
rs770726562 Chr.1: 155056913-155056913 on Build GRCh38
rs200526054 Chr.1: 155056968-155056968 on Build GRCh38
rs756882286 Chr.1: 155057019-155057019 on Build GRCh38
rs754264835 Chr.1: 155056997-155056997 on Build GRCh38
rs914867377 Chr.1: 155056853-155056853 on Build GRCh38
rs939137921 Chr.1: 155056920-155056920 on Build GRCh38
rs778768433 Chr.1: 155056906-155056907 on Build GRCh38
rs140070411 Chr.1: 155057016-155057016 on Build GRCh38
rs781728322 Chr.1: 155056980-155056980 on Build GRCh38
rs777186957 Chr.1: 155056994-155056994 on Build GRCh38
rs940287672 Chr.1: 155056899-155056899 on Build GRCh38
rs1049010288 Chr.1: 155057015-155057015 on Build GRCh38
rs572162097 Chr.1: 155056871-155056871 on Build GRCh38
rs776780564 Chr.1: 155056937-155056937 on Build GRCh38
rs989753925 Chr.1: 155056842-155056842 on Build GRCh38
rs772145588 Chr.1: 155056936-155056936 on Build GRCh38
rs897692518 Chr.1: 155056949-155056949 on Build GRCh38
rs745366484 Chr.1: 155056991-155056991 on Build GRCh38
rs368689211 Chr.1: 155056933-155056933 on Build GRCh38

View Full Product Details