Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 28524430-28524691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW2
Entrez Gene ID
152098
Gene Name
zinc finger CW-type and PWWP domain containing 2
Gene Aliases
ZCW2
UniGene
Hs.510635
Gene Chromosome Location
Chr.3: 28349149-28526366 on Build GRCh38
Transcripts
NM_001040432.3 NM_001324169.1 NM_001324170.1 NR_136708.1 XM_017005755.1 XM_017005756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2983341 c.939A>G p.A313A Chr.3: 28524556-28524556 on Build GRCh38
6582985 c.941G>C p.G314A Chr.3: 28524558-28524558 on Build GRCh38
2983346 c.1048G>C p.D350H Chr.3: 28524665-28524665 on Build GRCh38
6826018 c.1009G>T p.E337* Chr.3: 28524626-28524626 on Build GRCh38
2983343 c.1004C>A p.A335D Chr.3: 28524621-28524621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200677939 Chr.3: 28524635-28524635 on Build GRCh38
rs752882027 Chr.3: 28524658-28524658 on Build GRCh38
rs757264151 Chr.3: 28524484-28524484 on Build GRCh38
rs374257615 Chr.3: 28524597-28524597 on Build GRCh38
rs766856908 Chr.3: 28524649-28524649 on Build GRCh38
rs201046033 Chr.3: 28524589-28524589 on Build GRCh38
rs866557021 Chr.3: 28524623-28524623 on Build GRCh38
rs767591388 Chr.3: 28524656-28524656 on Build GRCh38
rs757031560 Chr.3: 28524596-28524596 on Build GRCh38
rs142286195 Chr.3: 28524547-28524547 on Build GRCh38
rs565462840 Chr.3: 28524613-28524613 on Build GRCh38
rs777104123 Chr.3: 28524545-28524545 on Build GRCh38
rs556427497 Chr.3: 28524480-28524480 on Build GRCh38
rs964578068 Chr.3: 28524580-28524580 on Build GRCh38
rs778681918 Chr.3: 28524494-28524494 on Build GRCh38
rs768413391 Chr.3: 28524538-28524538 on Build GRCh38
rs760547658 Chr.3: 28524556-28524557 on Build GRCh38
rs771724516 Chr.3: 28524502-28524502 on Build GRCh38
rs749019405 Chr.3: 28524482-28524482 on Build GRCh38
rs369567370 Chr.3: 28524579-28524579 on Build GRCh38
rs775076235 Chr.3: 28524513-28524513 on Build GRCh38
rs763518014 Chr.3: 28524648-28524648 on Build GRCh38
rs763391484 Chr.3: 28524576-28524576 on Build GRCh38
rs764207452 Chr.3: 28524478-28524478 on Build GRCh38
rs746730292 Chr.3: 28524522-28524522 on Build GRCh38
rs745774628 Chr.3: 28524501-28524501 on Build GRCh38
rs765348193 Chr.3: 28524561-28524561 on Build GRCh38
rs753704201 Chr.3: 28524591-28524591 on Build GRCh38
rs139764549 Chr.3: 28524610-28524610 on Build GRCh38
rs1055186367 Chr.3: 28524470-28524470 on Build GRCh38
rs755248175 Chr.3: 28524586-28524586 on Build GRCh38
rs1030281310 Chr.3: 28524592-28524592 on Build GRCh38
rs776181274 Chr.3: 28524626-28524626 on Build GRCh38
rs527953274 Chr.3: 28524630-28524630 on Build GRCh38
rs773613109 Chr.3: 28524569-28524569 on Build GRCh38
rs773556634 Chr.3: 28524645-28524645 on Build GRCh38
rs774459996 Chr.3: 28524651-28524651 on Build GRCh38
rs908410108 Chr.3: 28524551-28524551 on Build GRCh38
rs1000965192 Chr.3: 28524531-28524531 on Build GRCh38
rs768464189 Chr.3: 28524614-28524614 on Build GRCh38

View Full Product Details