Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132224346-132224585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
UniGene
Hs.469002
Gene Chromosome Location
Chr.10: 132207482-132332559 on Build GRCh38
Transcripts
NM_001318878.1 NM_001318879.1 NM_173575.3 NR_134911.1 XM_011539688.1 XM_011539690.2 XM_011539693.2 XM_011539694.1 XM_011539695.1 XM_011539696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5819844 c.901A>T p.N301Y Chr.10: 132224499-132224499 on Build GRCh38
3978254 c.967T>A p.F323I Chr.10: 132224472-132224472 on Build GRCh38
916345 c.904G>A p.A302T Chr.10: 132224496-132224496 on Build GRCh38
1195441 c.1018G>A p.A340T Chr.10: 132224421-132224421 on Build GRCh38
6410669 c.937G>A p.V313M Chr.10: 132224463-132224463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45527342 Chr.10: 132224527-132224527 on Build GRCh38
rs752906717 Chr.10: 132224479-132224479 on Build GRCh38
rs370546316 Chr.10: 132224557-132224557 on Build GRCh38
rs769354736 Chr.10: 132224426-132224426 on Build GRCh38
rs775169222 Chr.10: 132224434-132224434 on Build GRCh38
rs911874629 Chr.10: 132224470-132224470 on Build GRCh38
rs747529725 Chr.10: 132224399-132224399 on Build GRCh38
rs150696583 Chr.10: 132224422-132224422 on Build GRCh38
rs764387264 Chr.10: 132224565-132224565 on Build GRCh38
rs374005311 Chr.10: 132224531-132224531 on Build GRCh38
rs967904550 Chr.10: 132224453-132224453 on Build GRCh38
rs762661324 Chr.10: 132224436-132224436 on Build GRCh38
rs961819015 Chr.10: 132224430-132224430 on Build GRCh38
rs1004563825 Chr.10: 132224396-132224396 on Build GRCh38
rs763274459 Chr.10: 132224564-132224564 on Build GRCh38
rs572961386 Chr.10: 132224452-132224452 on Build GRCh38
rs745402229 Chr.10: 132224518-132224518 on Build GRCh38
rs753913139 Chr.10: 132224373-132224373 on Build GRCh38
rs990722829 Chr.10: 132224438-132224438 on Build GRCh38
rs774122560 Chr.10: 132224455-132224455 on Build GRCh38
rs377535352 Chr.10: 132224497-132224497 on Build GRCh38
rs755584279 Chr.10: 132224520-132224520 on Build GRCh38
rs201342384 Chr.10: 132224493-132224493 on Build GRCh38
rs758719022 Chr.10: 132224397-132224397 on Build GRCh38
rs780888116 Chr.10: 132224421-132224421 on Build GRCh38
rs202010879 Chr.10: 132224411-132224411 on Build GRCh38
rs775427698 Chr.10: 132224563-132224563 on Build GRCh38
rs200768058 Chr.10: 132224540-132224540 on Build GRCh38
rs574586225 Chr.10: 132224412-132224412 on Build GRCh38
rs185020377 Chr.10: 132224496-132224496 on Build GRCh38
rs149249276 Chr.10: 132224460-132224460 on Build GRCh38
rs986344003 Chr.10: 132224481-132224481 on Build GRCh38
rs1049664767 Chr.10: 132224539-132224539 on Build GRCh38
rs779145152 Chr.10: 132224377-132224377 on Build GRCh38
rs753031179 Chr.10: 132224378-132224378 on Build GRCh38
rs772025463 Chr.10: 132224546-132224546 on Build GRCh38
rs925786798 Chr.10: 132224561-132224561 on Build GRCh38
rs554327105 Chr.10: 132224371-132224371 on Build GRCh38
rs749030656 Chr.10: 132224529-132224529 on Build GRCh38
rs139671041 Chr.10: 132224457-132224457 on Build GRCh38
rs561459094 Chr.10: 132224541-132224541 on Build GRCh38
rs577074026 Chr.10: 132224463-132224463 on Build GRCh38
rs534089521 Chr.10: 132224440-132224440 on Build GRCh38
rs141959046 Chr.10: 132224464-132224464 on Build GRCh38
rs896145293 Chr.10: 132224558-132224558 on Build GRCh38
rs776925668 Chr.10: 132224458-132224458 on Build GRCh38
rs765414370 Chr.10: 132224562-132224562 on Build GRCh38
rs766507175 Chr.10: 132224368-132224368 on Build GRCh38
rs748305044 Chr.10: 132224374-132224374 on Build GRCh38
rs377648670 Chr.10: 132224556-132224556 on Build GRCh38
rs886152262 Chr.10: 132224388-132224388 on Build GRCh38
rs781746681 Chr.10: 132224503-132224503 on Build GRCh38
rs545038961 Chr.10: 132224494-132224494 on Build GRCh38
rs547176886 Chr.10: 132224560-132224560 on Build GRCh38

View Full Product Details