Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78803315-78803586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP36
Entrez Gene ID
57602
Gene Name
ubiquitin specific peptidase 36
Gene Aliases
DUB1
UniGene
Hs.464243
Gene Chromosome Location
Chr.17: 78787381-78840898 on Build GRCh38
Transcripts
NM_001321291.1 XM_005257542.2 XM_005257543.2 XM_011525065.1 XM_011525066.2 XM_011525067.2 XM_011525068.1 XM_011525069.2 XM_011525070.1 XM_011525071.2 XM_017024900.1 XM_017024901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
340482 c.2758G>C p.A920P Chr.17: 78803437-78803437 on Build GRCh38
4539841 c.2744G>A p.R915Q Chr.17: 78803451-78803451 on Build GRCh38
6818648 c.2727C>T p.H909H Chr.17: 78803468-78803468 on Build GRCh38
6322215 c.2659C>A p.R887R Chr.17: 78803536-78803536 on Build GRCh38
1494097 c.2655C>T p.V885V Chr.17: 78803540-78803540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758907001 Chr.17: 78803484-78803489 on Build GRCh38
rs760166513 Chr.17: 78803348-78803349 on Build GRCh38
rs764209207 Chr.17: 78803366-78803366 on Build GRCh38
rs563529475 Chr.17: 78803462-78803462 on Build GRCh38
rs764331946 Chr.17: 78803471-78803471 on Build GRCh38
rs758215486 Chr.17: 78803510-78803510 on Build GRCh38
rs758375326 Chr.17: 78803405-78803405 on Build GRCh38
rs769146023 Chr.17: 78803538-78803538 on Build GRCh38
rs751504294 Chr.17: 78803521-78803521 on Build GRCh38
rs763643610 Chr.17: 78803358-78803358 on Build GRCh38
rs569678575 Chr.17: 78803348-78803348 on Build GRCh38
rs375588270 Chr.17: 78803346-78803346 on Build GRCh38
rs545764041 Chr.17: 78803466-78803466 on Build GRCh38
rs200106890 Chr.17: 78803476-78803476 on Build GRCh38
rs577196991 Chr.17: 78803465-78803465 on Build GRCh38
rs753214330 Chr.17: 78803359-78803359 on Build GRCh38
rs372509099 Chr.17: 78803391-78803391 on Build GRCh38
rs140332801 Chr.17: 78803546-78803546 on Build GRCh38
rs944383596 Chr.17: 78803424-78803424 on Build GRCh38
rs774617830 Chr.17: 78803456-78803456 on Build GRCh38
rs140617093 Chr.17: 78803414-78803414 on Build GRCh38
rs779767309 Chr.17: 78803519-78803519 on Build GRCh38
rs754008998 Chr.17: 78803369-78803369 on Build GRCh38
rs765471675 Chr.17: 78803481-78803481 on Build GRCh38
rs750573740 Chr.17: 78803493-78803493 on Build GRCh38
rs144517101 Chr.17: 78803452-78803452 on Build GRCh38
rs766657162 Chr.17: 78803345-78803345 on Build GRCh38
rs750978736 Chr.17: 78803476-78803482 on Build GRCh38
rs776054280 Chr.17: 78803436-78803436 on Build GRCh38
rs1027241768 Chr.17: 78803408-78803408 on Build GRCh38
rs61760231 Chr.17: 78803535-78803535 on Build GRCh38
rs776618341 Chr.17: 78803561-78803561 on Build GRCh38
rs771709549 Chr.17: 78803555-78803555 on Build GRCh38
rs745657752 Chr.17: 78803392-78803392 on Build GRCh38
rs140856412 Chr.17: 78803467-78803467 on Build GRCh38
rs777501860 Chr.17: 78803540-78803540 on Build GRCh38
rs748890796 Chr.17: 78803544-78803544 on Build GRCh38
rs768169484 Chr.17: 78803431-78803431 on Build GRCh38
rs747662867 Chr.17: 78803536-78803536 on Build GRCh38
rs949899269 Chr.17: 78803340-78803340 on Build GRCh38
rs574545654 Chr.17: 78803451-78803451 on Build GRCh38
rs780000218 Chr.17: 78803413-78803413 on Build GRCh38
rs951591556 Chr.17: 78803356-78803356 on Build GRCh38
rs757493080 Chr.17: 78803372-78803372 on Build GRCh38
rs747771918 Chr.17: 78803545-78803545 on Build GRCh38
rs773848703 Chr.17: 78803472-78803472 on Build GRCh38
rs368552550 Chr.17: 78803360-78803360 on Build GRCh38
rs537842306 Chr.17: 78803468-78803468 on Build GRCh38

View Full Product Details