Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100374955-100375192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOMM70
Entrez Gene ID
9868
Gene Name
translocase of outer mitochondrial membrane 70
Gene Aliases
TOMM70A, Tom70
UniGene
Hs.227253
Gene Chromosome Location
Chr.3: 100363459-100401398 on Build GRCh38
Transcripts
NM_014820.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726658 c.1097G>T p.R366L Chr.3: 100375148-100375148 on Build GRCh38
6280423 c.1093-1G>T p.? Chr.3: 100375153-100375153 on Build GRCh38
726659 c.1220G>T p.R407L Chr.3: 100375025-100375025 on Build GRCh38
4169665 c.1099G>A p.A367T Chr.3: 100375146-100375146 on Build GRCh38
3584507 c.1118G>A p.R373K Chr.3: 100375127-100375127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140101095 Chr.3: 100375052-100375052 on Build GRCh38
rs766516311 Chr.3: 100375086-100375086 on Build GRCh38
rs535327192 Chr.3: 100375027-100375027 on Build GRCh38
rs753907950 Chr.3: 100375095-100375095 on Build GRCh38
rs777630377 Chr.3: 100375100-100375100 on Build GRCh38
rs754836393 Chr.3: 100375160-100375161 on Build GRCh38
rs368337713 Chr.3: 100374992-100374992 on Build GRCh38
rs770837618 Chr.3: 100374987-100374987 on Build GRCh38
rs748804437 Chr.3: 100375148-100375148 on Build GRCh38
rs1029436274 Chr.3: 100375026-100375026 on Build GRCh38
rs774841593 Chr.3: 100375158-100375158 on Build GRCh38
rs767798979 Chr.3: 100375059-100375059 on Build GRCh38
rs202011920 Chr.3: 100375137-100375137 on Build GRCh38
rs945536542 Chr.3: 100375049-100375049 on Build GRCh38
rs769090895 Chr.3: 100375048-100375048 on Build GRCh38
rs762230068 Chr.3: 100375056-100375056 on Build GRCh38
rs750653539 Chr.3: 100375060-100375060 on Build GRCh38
rs757122794 Chr.3: 100375123-100375123 on Build GRCh38
rs755066122 Chr.3: 100375098-100375098 on Build GRCh38
rs144929730 Chr.3: 100375046-100375046 on Build GRCh38
rs768203450 Chr.3: 100375155-100375155 on Build GRCh38
rs890442926 Chr.3: 100375047-100375047 on Build GRCh38
rs774822266 Chr.3: 100375004-100375004 on Build GRCh38
rs369224783 Chr.3: 100375009-100375009 on Build GRCh38
rs745545730 Chr.3: 100375135-100375135 on Build GRCh38
rs779733579 Chr.3: 100375139-100375139 on Build GRCh38
rs781084451 Chr.3: 100375129-100375129 on Build GRCh38
rs751540536 Chr.3: 100375119-100375119 on Build GRCh38
rs751330736 Chr.3: 100375008-100375009 on Build GRCh38
rs760841775 Chr.3: 100375081-100375081 on Build GRCh38
rs372893004 Chr.3: 100374995-100374995 on Build GRCh38
rs746851104 Chr.3: 100374984-100374984 on Build GRCh38
rs60463428 Chr.3: 100374997-100374997 on Build GRCh38

View Full Product Details