Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38741610-38741837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH8
Entrez Gene ID
1769
Gene Name
dynein axonemal heavy chain 8
Gene Aliases
ATPase, hdhc9
UniGene
Hs.520106
Gene Chromosome Location
Chr.6: 38715329-39030798 on Build GRCh38
Transcripts
NM_001206927.1 XM_011514318.2 XM_011514319.2 XM_011514320.2 XM_011514321.1 XM_017010325.1 XM_017010326.1 XM_017010327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297585 c.1117-8G>T p.? Chr.6: 38741703-38741703 on Build GRCh38
1734445 c.541C>T p.R181C Chr.6: 38741786-38741786 on Build GRCh38
6847811 c.529G>T p.E177* Chr.6: 38741774-38741774 on Build GRCh38
6646833 c.1128G>A p.E376E Chr.6: 38741722-38741722 on Build GRCh38
6529783 c.1193G>C p.R398P Chr.6: 38741787-38741787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773740103 Chr.6: 38741682-38741682 on Build GRCh38
rs763602579 Chr.6: 38741678-38741678 on Build GRCh38
rs765653307 Chr.6: 38741719-38741719 on Build GRCh38
rs370751861 Chr.6: 38741786-38741786 on Build GRCh38
rs924945215 Chr.6: 38741695-38741695 on Build GRCh38
rs552179604 Chr.6: 38741727-38741727 on Build GRCh38
rs374073806 Chr.6: 38741790-38741790 on Build GRCh38
rs773062035 Chr.6: 38741751-38741753 on Build GRCh38
rs760373783 Chr.6: 38741805-38741805 on Build GRCh38
rs936352893 Chr.6: 38741707-38741707 on Build GRCh38
rs867794988 Chr.6: 38741759-38741759 on Build GRCh38
rs761488169 Chr.6: 38741691-38741691 on Build GRCh38
rs767181931 Chr.6: 38741708-38741708 on Build GRCh38
rs978443907 Chr.6: 38741681-38741681 on Build GRCh38
rs771614521 Chr.6: 38741802-38741802 on Build GRCh38
rs768349241 Chr.6: 38741663-38741665 on Build GRCh38
rs751620349 Chr.6: 38741741-38741741 on Build GRCh38
rs758840030 Chr.6: 38741730-38741730 on Build GRCh38
rs1051264232 Chr.6: 38741787-38741787 on Build GRCh38
rs779727583 Chr.6: 38741771-38741771 on Build GRCh38
rs201355783 Chr.6: 38741776-38741776 on Build GRCh38
rs774020549 Chr.6: 38741672-38741673 on Build GRCh38
rs750127269 Chr.6: 38741709-38741709 on Build GRCh38
rs532383670 Chr.6: 38741642-38741642 on Build GRCh38
rs141326848 Chr.6: 38741757-38741757 on Build GRCh38
rs567656319 Chr.6: 38741687-38741688 on Build GRCh38
rs772850492 Chr.6: 38741804-38741804 on Build GRCh38
rs777839170 Chr.6: 38741736-38741736 on Build GRCh38
rs771580014 Chr.6: 38741717-38741720 on Build GRCh38
rs377401999 Chr.6: 38741749-38741749 on Build GRCh38
rs559609199 Chr.6: 38741742-38741742 on Build GRCh38
rs768508741 Chr.6: 38741778-38741778 on Build GRCh38
rs759866696 Chr.6: 38741718-38741718 on Build GRCh38

View Full Product Details