Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 101543326-101543555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL2
Entrez Gene ID
79977
Gene Name
grainyhead like transcription factor 2
Gene Aliases
BOM, DFNA28, ECTDS, TFCP2L3
UniGene
Hs.661088
Gene Chromosome Location
Chr.8: 101492440-101678105 on Build GRCh38
Transcripts
NM_024915.3 XM_011517305.2 XM_011517306.2 XM_011517307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6179246 c.120G>T p.L40F Chr.8: 101543340-101543340 on Build GRCh38
4523553 c.121G>A p.E41K Chr.8: 101543341-101543341 on Build GRCh38
7157414 c.192C>T p.L64L Chr.8: 101543412-101543412 on Build GRCh38
4978938 c.184G>C p.A62P Chr.8: 101543404-101543404 on Build GRCh38
4523888 c.123G>A p.E41E Chr.8: 101543343-101543343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747378549 Chr.8: 101543522-101543522 on Build GRCh38
rs769228709 Chr.8: 101543349-101543349 on Build GRCh38
rs754709953 Chr.8: 101543480-101543480 on Build GRCh38
rs1019441261 Chr.8: 101543421-101543421 on Build GRCh38
rs746434474 Chr.8: 101543412-101543412 on Build GRCh38
rs748743115 Chr.8: 101543518-101543518 on Build GRCh38
rs202225242 Chr.8: 101543476-101543476 on Build GRCh38
rs765356115 Chr.8: 101543465-101543465 on Build GRCh38
rs376505205 Chr.8: 101543444-101543444 on Build GRCh38
rs199992903 Chr.8: 101543443-101543443 on Build GRCh38
rs777381691 Chr.8: 101543513-101543513 on Build GRCh38
rs928093748 Chr.8: 101543459-101543459 on Build GRCh38
rs751433224 Chr.8: 101543478-101543478 on Build GRCh38
rs762012342 Chr.8: 101543455-101543455 on Build GRCh38
rs766225382 Chr.8: 101543477-101543477 on Build GRCh38
rs750449679 Chr.8: 101543471-101543471 on Build GRCh38
rs776923689 Chr.8: 101543449-101543449 on Build GRCh38
rs774752882 Chr.8: 101543403-101543403 on Build GRCh38
rs975289815 Chr.8: 101543441-101543441 on Build GRCh38
rs775989224 Chr.8: 101543436-101543436 on Build GRCh38
rs772426012 Chr.8: 101543423-101543423 on Build GRCh38
rs747761110 Chr.8: 101543496-101543496 on Build GRCh38
rs769099387 Chr.8: 101543526-101543526 on Build GRCh38
rs993904280 Chr.8: 101543356-101543356 on Build GRCh38
rs772692559 Chr.8: 101543520-101543520 on Build GRCh38
rs764419988 Chr.8: 101543445-101543445 on Build GRCh38
rs775799518 Chr.8: 101543521-101543521 on Build GRCh38
rs746625344 Chr.8: 101543507-101543507 on Build GRCh38
rs780828381 Chr.8: 101543484-101543484 on Build GRCh38

View Full Product Details