Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70449689-70449944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_021120.3 XM_006724625.2 XM_006724626.2 XM_011530883.1

SNP Detail

SNP ID Chromosome Location
rs778285381 Chr.X: 70449759-70449759 on Build GRCh38
rs746761243 Chr.X: 70449816-70449816 on Build GRCh38
rs766946010 Chr.X: 70449895-70449895 on Build GRCh38
rs773102254 Chr.X: 70449790-70449790 on Build GRCh38
rs150996616 Chr.X: 70449732-70449732 on Build GRCh38
rs953081519 Chr.X: 70449786-70449786 on Build GRCh38
rs750963382 Chr.X: 70449789-70449789 on Build GRCh38
rs975881791 Chr.X: 70449740-70449740 on Build GRCh38
rs979467868 Chr.X: 70449798-70449798 on Build GRCh38
rs754423267 Chr.X: 70449753-70449753 on Build GRCh38
rs750357993 Chr.X: 70449727-70449727 on Build GRCh38
rs758870192 Chr.X: 70449818-70449818 on Build GRCh38
rs369819281 Chr.X: 70449735-70449735 on Build GRCh38
rs759695364 Chr.X: 70449855-70449855 on Build GRCh38
rs794727648 Chr.X: 70449726-70449726 on Build GRCh38
rs748814438 Chr.X: 70449749-70449749 on Build GRCh38
rs1052288465 Chr.X: 70449868-70449868 on Build GRCh38
rs747897416 Chr.X: 70449762-70449762 on Build GRCh38
rs902115508 Chr.X: 70449834-70449834 on Build GRCh38
rs765478924 Chr.X: 70449880-70449880 on Build GRCh38
rs984580660 Chr.X: 70449752-70449752 on Build GRCh38
rs763284789 Chr.X: 70449897-70449897 on Build GRCh38
rs751846954 Chr.X: 70449731-70449731 on Build GRCh38
rs924946746 Chr.X: 70449819-70449819 on Build GRCh38
rs200883696 Chr.X: 70449729-70449729 on Build GRCh38
rs142606535 Chr.X: 70449708-70449708 on Build GRCh38

View Full Product Details