Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43453271-43453520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYI
Entrez Gene ID
81888
Gene Name
hydroxypyruvate isomerase (putative)
Gene Aliases
HT036
UniGene
Hs.709864
Gene Chromosome Location
Chr.1: 43450607-43454241 on Build GRCh38
Transcripts
NM_001190880.2 NM_001243526.1 NM_031207.5 XM_005271239.4 XM_005271240.4 XM_006710937.3 XM_011542222.2 NM_015284.3

Gene Symbol
SZT2
Entrez Gene ID
23334
Gene Name
seizure threshold 2 homolog (mouse)
Gene Aliases
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B
UniGene
Hs.643560
Gene Chromosome Location
Chr.1: 43389884-43454247 on Build GRCh38
Transcripts
NM_001190880.2 NM_001243526.1 NM_031207.5 XM_005271239.4 XM_005271240.4 XM_006710937.3 XM_011542222.2 NM_015284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242360 c.226C>A p.L76M Chr.1: 43453471-43453471 on Build GRCh38
909863 c.210G>T p.E70D Chr.1: 43453487-43453487 on Build GRCh38
7300948 c.21delC p.R9fs*19 Chr.1: 43453457-43453457 on Build GRCh38
4143987 c.287A>G p.Y96C Chr.1: 43453410-43453410 on Build GRCh38
1210198 c.75A>C p.K25N Chr.1: 43453403-43453403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755989872 Chr.1: 43453456-43453456 on Build GRCh38
rs867461845 Chr.1: 43453414-43453414 on Build GRCh38
rs910061927 Chr.1: 43453297-43453297 on Build GRCh38
rs975683949 Chr.1: 43453381-43453381 on Build GRCh38
rs762168706 Chr.1: 43453423-43453423 on Build GRCh38
rs758615239 Chr.1: 43453429-43453429 on Build GRCh38
rs762498837 Chr.1: 43453413-43453413 on Build GRCh38
rs972831214 Chr.1: 43453493-43453493 on Build GRCh38
rs545302537 Chr.1: 43453406-43453406 on Build GRCh38
rs748934658 Chr.1: 43453463-43453463 on Build GRCh38
rs866325841 Chr.1: 43453444-43453444 on Build GRCh38
rs777772412 Chr.1: 43453460-43453460 on Build GRCh38
rs765384919 Chr.1: 43453427-43453427 on Build GRCh38
rs764529011 Chr.1: 43453420-43453420 on Build GRCh38
rs141158872 Chr.1: 43453395-43453395 on Build GRCh38
rs905464006 Chr.1: 43453487-43453487 on Build GRCh38
rs572238649 Chr.1: 43453428-43453428 on Build GRCh38
rs781311591 Chr.1: 43453447-43453447 on Build GRCh38
rs1037542115 Chr.1: 43453344-43453344 on Build GRCh38
rs965683182 Chr.1: 43453377-43453377 on Build GRCh38
rs750435547 Chr.1: 43453369-43453369 on Build GRCh38
rs556116001 Chr.1: 43453400-43453400 on Build GRCh38
rs747888724 Chr.1: 43453455-43453455 on Build GRCh38
rs773770463 Chr.1: 43453419-43453419 on Build GRCh38
rs372269751 Chr.1: 43453357-43453358 on Build GRCh38
rs751640782 Chr.1: 43453433-43453433 on Build GRCh38
rs983639138 Chr.1: 43453298-43453298 on Build GRCh38
rs769534662 Chr.1: 43453409-43453409 on Build GRCh38
rs755702385 Chr.1: 43453476-43453477 on Build GRCh38
rs935171945 Chr.1: 43453415-43453415 on Build GRCh38
rs779135534 Chr.1: 43453366-43453366 on Build GRCh38
rs1013599992 Chr.1: 43453481-43453481 on Build GRCh38
rs377473419 Chr.1: 43453356-43453356 on Build GRCh38
rs375922579 Chr.1: 43453360-43453360 on Build GRCh38
rs371668641 Chr.1: 43453480-43453480 on Build GRCh38
rs142369206 Chr.1: 43453410-43453410 on Build GRCh38
rs137998671 Chr.1: 43453402-43453402 on Build GRCh38
rs754897289 Chr.1: 43453435-43453435 on Build GRCh38
rs369407159 Chr.1: 43453492-43453492 on Build GRCh38
rs372316003 Chr.1: 43453349-43453349 on Build GRCh38
rs773859072 Chr.1: 43453484-43453484 on Build GRCh38
rs377170811 Chr.1: 43453466-43453466 on Build GRCh38
rs764485556 Chr.1: 43453359-43453359 on Build GRCh38
rs1045060861 Chr.1: 43453486-43453486 on Build GRCh38
rs866645901 Chr.1: 43453462-43453462 on Build GRCh38
rs74598418 Chr.1: 43453302-43453302 on Build GRCh38
rs896532689 Chr.1: 43453469-43453469 on Build GRCh38
rs369028134 Chr.1: 43453383-43453383 on Build GRCh38
rs372637553 Chr.1: 43453397-43453397 on Build GRCh38
rs770465127 Chr.1: 43453417-43453417 on Build GRCh38
rs766452814 Chr.1: 43453432-43453432 on Build GRCh38
rs776939158 Chr.1: 43453422-43453422 on Build GRCh38
rs906855183 Chr.1: 43453443-43453443 on Build GRCh38
rs1002975255 Chr.1: 43453446-43453446 on Build GRCh38
rs1035274867 Chr.1: 43453452-43453452 on Build GRCh38
rs757419989 Chr.1: 43453363-43453363 on Build GRCh38

View Full Product Details