Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 36892276-36892501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIS2
Entrez Gene ID
4212
Gene Name
Meis homeobox 2
Gene Aliases
HsT18361, MRG1
UniGene
Hs.510989
Gene Chromosome Location
Chr.15: 36891021-37101299 on Build GRCh38
Transcripts
NM_001220482.1 NM_002399.3 NM_170674.4 NM_170675.4 NM_170676.4 NM_170677.4 NM_172315.2 NM_172316.2 NR_051953.1 XM_006720523.1 XM_006720524.1 XM_006720525.1 XM_006720526.2 XM_006720527.2 XM_006720529.2 XM_011521591.2 XM_017022205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707989 c.1280C>T p.S427L Chr.15: 36892327-36892327 on Build GRCh38
4435402 c.1235T>G p.M412R Chr.15: 36892372-36892372 on Build GRCh38
433750 c.1246C>T p.P416S Chr.15: 36892361-36892361 on Build GRCh38
7181037 c.1169A>G p.D390G Chr.15: 36892438-36892438 on Build GRCh38
240653 c.1270_1287del18 p.P424_L429delPMHSYL Chr.15: 36892320-36892337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199728455 Chr.15: 36892344-36892344 on Build GRCh38
rs57860578 Chr.15: 36892374-36892374 on Build GRCh38
rs142878520 Chr.15: 36892426-36892426 on Build GRCh38
rs775995092 Chr.15: 36892455-36892455 on Build GRCh38
rs987970014 Chr.15: 36892423-36892423 on Build GRCh38
rs753026080 Chr.15: 36892340-36892340 on Build GRCh38
rs749988263 Chr.15: 36892466-36892466 on Build GRCh38
rs773532150 Chr.15: 36892306-36892306 on Build GRCh38
rs760021973 Chr.15: 36892334-36892334 on Build GRCh38
rs764528465 Chr.15: 36892464-36892464 on Build GRCh38
rs781319540 Chr.15: 36892415-36892415 on Build GRCh38
rs759650078 Chr.15: 36892450-36892450 on Build GRCh38
rs943819281 Chr.15: 36892465-36892465 on Build GRCh38
rs758059682 Chr.15: 36892467-36892467 on Build GRCh38
rs767985608 Chr.15: 36892451-36892451 on Build GRCh38
rs779632587 Chr.15: 36892364-36892364 on Build GRCh38
rs774662563 Chr.15: 36892322-36892322 on Build GRCh38
rs560301639 Chr.15: 36892420-36892420 on Build GRCh38
rs549296314 Chr.15: 36892470-36892470 on Build GRCh38
rs768077389 Chr.15: 36892339-36892339 on Build GRCh38
rs533167442 Chr.15: 36892321-36892321 on Build GRCh38
rs930989017 Chr.15: 36892414-36892414 on Build GRCh38
rs746560276 Chr.15: 36892369-36892369 on Build GRCh38
rs754963373 Chr.15: 36892408-36892408 on Build GRCh38
rs374458760 Chr.15: 36892298-36892298 on Build GRCh38
rs971258285 Chr.15: 36892308-36892308 on Build GRCh38
rs151055354 Chr.15: 36892433-36892433 on Build GRCh38
rs144548752 Chr.15: 36892345-36892345 on Build GRCh38
rs1032019811 Chr.15: 36892363-36892363 on Build GRCh38
rs760991030 Chr.15: 36892462-36892462 on Build GRCh38
rs774482745 Chr.15: 36892447-36892447 on Build GRCh38
rs749536909 Chr.15: 36892434-36892434 on Build GRCh38
rs531139958 Chr.15: 36892441-36892441 on Build GRCh38
rs113747758 Chr.15: 36892393-36892393 on Build GRCh38
rs956554759 Chr.15: 36892390-36892390 on Build GRCh38
rs757783328 Chr.15: 36892354-36892354 on Build GRCh38
rs148444214 Chr.15: 36892353-36892353 on Build GRCh38
rs770111561 Chr.15: 36892303-36892303 on Build GRCh38
rs762901201 Chr.15: 36892319-36892319 on Build GRCh38
rs141054432 Chr.15: 36892295-36892295 on Build GRCh38

View Full Product Details