Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33204883-33205126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425884 c.128T>C p.L43P Chr.6: 33204977-33204977 on Build GRCh38
3994877 c.195C>T p.P65P Chr.6: 33205044-33205044 on Build GRCh38
5734247 c.150G>C p.E50D Chr.6: 33204999-33204999 on Build GRCh38
4886974 c.235G>T p.A79S Chr.6: 33205084-33205084 on Build GRCh38
6682386 c.239G>T p.R80M Chr.6: 33205088-33205088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754817548 Chr.6: 33205054-33205054 on Build GRCh38
rs975455369 Chr.6: 33205043-33205043 on Build GRCh38
rs111493353 Chr.6: 33204978-33204978 on Build GRCh38
rs768841679 Chr.6: 33204981-33204981 on Build GRCh38
rs1048747696 Chr.6: 33205048-33205048 on Build GRCh38
rs748042744 Chr.6: 33204942-33204942 on Build GRCh38
rs749983676 Chr.6: 33205014-33205014 on Build GRCh38
rs770107202 Chr.6: 33204972-33204972 on Build GRCh38
rs762264687 Chr.6: 33204995-33204995 on Build GRCh38
rs777835548 Chr.6: 33205102-33205102 on Build GRCh38
rs956937028 Chr.6: 33204909-33204909 on Build GRCh38
rs754724398 Chr.6: 33204938-33204938 on Build GRCh38
rs528195394 Chr.6: 33205074-33205074 on Build GRCh38
rs1032990828 Chr.6: 33205002-33205002 on Build GRCh38
rs771158268 Chr.6: 33204945-33204945 on Build GRCh38
rs1036445906 Chr.6: 33204906-33204906 on Build GRCh38
rs762802518 Chr.6: 33205022-33205023 on Build GRCh38
rs766344422 Chr.6: 33204976-33204976 on Build GRCh38
rs760163346 Chr.6: 33205034-33205034 on Build GRCh38
rs953455532 Chr.6: 33204937-33204937 on Build GRCh38
rs898887165 Chr.6: 33204926-33204926 on Build GRCh38
rs758449941 Chr.6: 33205092-33205092 on Build GRCh38
rs563903455 Chr.6: 33204989-33204989 on Build GRCh38
rs202140618 Chr.6: 33204928-33204928 on Build GRCh38
rs915354788 Chr.6: 33204932-33204932 on Build GRCh38
rs745915980 Chr.6: 33204961-33204961 on Build GRCh38
rs919493169 Chr.6: 33205010-33205010 on Build GRCh38
rs774942115 Chr.6: 33204990-33204990 on Build GRCh38
rs367755157 Chr.6: 33205007-33205007 on Build GRCh38
rs370759993 Chr.6: 33204921-33204921 on Build GRCh38
rs569282496 Chr.6: 33204939-33204939 on Build GRCh38
rs368394952 Chr.6: 33204960-33204960 on Build GRCh38
rs894301835 Chr.6: 33204956-33204956 on Build GRCh38
rs766121236 Chr.6: 33205036-33205036 on Build GRCh38
rs752713870 Chr.6: 33205075-33205075 on Build GRCh38
rs930329403 Chr.6: 33205041-33205041 on Build GRCh38
rs753364027 Chr.6: 33205046-33205046 on Build GRCh38
rs763890055 Chr.6: 33205053-33205064 on Build GRCh38

View Full Product Details