Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157382169-157382394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB6
Entrez Gene ID
10049
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
UniGene
Hs.490745
Gene Chromosome Location
Chr.7: 157337004-157417439 on Build GRCh38
Transcripts
NM_005494.2 NM_058246.3 XM_005249515.2 XM_005249516.2 XM_006715823.1 XM_011515704.1 XM_017011633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580887 c.446C>T p.S149F Chr.7: 157382345-157382345 on Build GRCh38
3305952 c.388C>T p.R130* Chr.7: 157382287-157382287 on Build GRCh38
3305956 c.440T>A p.F147Y Chr.7: 157382339-157382339 on Build GRCh38
1699649 c.443C>T p.P148L Chr.7: 157382342-157382342 on Build GRCh38
4958908 c.407G>A p.G136E Chr.7: 157382306-157382306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003658028 Chr.7: 157382332-157382332 on Build GRCh38
rs143051731 Chr.7: 157382316-157382316 on Build GRCh38
rs757767434 Chr.7: 157382327-157382327 on Build GRCh38
rs776104560 Chr.7: 157382216-157382216 on Build GRCh38
rs748020003 Chr.7: 157382289-157382289 on Build GRCh38
rs375208186 Chr.7: 157382264-157382264 on Build GRCh38
rs763581069 Chr.7: 157382218-157382218 on Build GRCh38
rs764748999 Chr.7: 157382222-157382222 on Build GRCh38
rs989242880 Chr.7: 157382348-157382348 on Build GRCh38
rs899358771 Chr.7: 157382299-157382299 on Build GRCh38
rs552218882 Chr.7: 157382206-157382206 on Build GRCh38
rs368565876 Chr.7: 157382276-157382276 on Build GRCh38
rs149027078 Chr.7: 157382309-157382309 on Build GRCh38
rs772308715 Chr.7: 157382214-157382215 on Build GRCh38
rs368036062 Chr.7: 157382232-157382232 on Build GRCh38
rs746162016 Chr.7: 157382208-157382208 on Build GRCh38
rs755794590 Chr.7: 157382239-157382239 on Build GRCh38
rs769285457 Chr.7: 157382311-157382311 on Build GRCh38
rs201051578 Chr.7: 157382272-157382272 on Build GRCh38
rs777224069 Chr.7: 157382291-157382291 on Build GRCh38
rs754885609 Chr.7: 157382269-157382269 on Build GRCh38
rs774544702 Chr.7: 157382227-157382227 on Build GRCh38
rs200698204 Chr.7: 157382256-157382256 on Build GRCh38
rs775051801 Chr.7: 157382312-157382312 on Build GRCh38
rs747600463 Chr.7: 157382280-157382280 on Build GRCh38
rs771621070 Chr.7: 157382203-157382203 on Build GRCh38
rs754826358 Chr.7: 157382343-157382343 on Build GRCh38
rs112310856 Chr.7: 157382337-157382337 on Build GRCh38
rs544075361 Chr.7: 157382207-157382207 on Build GRCh38
rs762017341 Chr.7: 157382228-157382228 on Build GRCh38
rs770199326 Chr.7: 157382213-157382213 on Build GRCh38
rs773234613 Chr.7: 157382361-157382363 on Build GRCh38
rs371893136 Chr.7: 157382263-157382263 on Build GRCh38
rs370712357 Chr.7: 157382223-157382223 on Build GRCh38
rs778861039 Chr.7: 157382270-157382270 on Build GRCh38
rs150709673 Chr.7: 157382328-157382328 on Build GRCh38
rs373837766 Chr.7: 157382226-157382226 on Build GRCh38
rs766637787 Chr.7: 157382331-157382331 on Build GRCh38
rs746706599 Chr.7: 157382302-157382302 on Build GRCh38
rs765102730 Chr.7: 157382358-157382358 on Build GRCh38
rs749714667 Chr.7: 157382310-157382310 on Build GRCh38
rs772121051 Chr.7: 157382303-157382303 on Build GRCh38
rs757941397 Chr.7: 157382285-157382285 on Build GRCh38

View Full Product Details