Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107353333-107353604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWF19L2
Entrez Gene ID
143884
Gene Name
CWF19-like 2, cell cycle control (S. pombe)
Gene Aliases
-
UniGene
Hs.212140
Gene Chromosome Location
Chr.11: 107326345-107457846 on Build GRCh38
Transcripts
NM_152434.2 XM_017017239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363383 c.1594A>C p.K532Q Chr.11: 107353553-107353553 on Build GRCh38
4476250 c.1591C>T p.P531S Chr.11: 107353556-107353556 on Build GRCh38
6638395 c.2034T>G p.C678W Chr.11: 107353575-107353575 on Build GRCh38
7145776 c.2017G>A p.E673K Chr.11: 107353592-107353592 on Build GRCh38
273775 c.1560_1561insA p.C521fs*6 Chr.11: 107353586-107353587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374724322 Chr.11: 107353489-107353489 on Build GRCh38
rs774691772 Chr.11: 107353573-107353573 on Build GRCh38
rs548600043 Chr.11: 107353482-107353482 on Build GRCh38
rs758014191 Chr.11: 107353510-107353510 on Build GRCh38
rs1054954679 Chr.11: 107353447-107353447 on Build GRCh38
rs750528079 Chr.11: 107353568-107353568 on Build GRCh38
rs937750992 Chr.11: 107353417-107353417 on Build GRCh38
rs759947345 Chr.11: 107353487-107353487 on Build GRCh38
rs761663112 Chr.11: 107353524-107353524 on Build GRCh38
rs751758513 Chr.11: 107353377-107353377 on Build GRCh38
rs908921797 Chr.11: 107353413-107353413 on Build GRCh38
rs918916312 Chr.11: 107353374-107353374 on Build GRCh38
rs199938531 Chr.11: 107353493-107353493 on Build GRCh38
rs756890162 Chr.11: 107353497-107353497 on Build GRCh38
rs956146111 Chr.11: 107353579-107353579 on Build GRCh38
rs761921764 Chr.11: 107353387-107353387 on Build GRCh38
rs779719294 Chr.11: 107353516-107353516 on Build GRCh38
rs749894980 Chr.11: 107353500-107353500 on Build GRCh38
rs4576794 Chr.11: 107353499-107353499 on Build GRCh38
rs781166288 Chr.11: 107353546-107353546 on Build GRCh38
rs759786778 Chr.11: 107353475-107353475 on Build GRCh38
rs375149241 Chr.11: 107353472-107353473 on Build GRCh38
rs768184317 Chr.11: 107353488-107353488 on Build GRCh38
rs928935421 Chr.11: 107353386-107353386 on Build GRCh38
rs748074300 Chr.11: 107353551-107353551 on Build GRCh38
rs760171636 Chr.11: 107353395-107353395 on Build GRCh38
rs767816267 Chr.11: 107353475-107353476 on Build GRCh38
rs896346584 Chr.11: 107353458-107353458 on Build GRCh38
rs929995578 Chr.11: 107353561-107353561 on Build GRCh38
rs768418998 Chr.11: 107353543-107353543 on Build GRCh38
rs139979724 Chr.11: 107353556-107353556 on Build GRCh38
rs1013492431 Chr.11: 107353464-107353464 on Build GRCh38
rs552668912 Chr.11: 107353537-107353537 on Build GRCh38
rs959672225 Chr.11: 107353437-107353437 on Build GRCh38
rs146504700 Chr.11: 107353553-107353553 on Build GRCh38
rs199558505 Chr.11: 107353569-107353569 on Build GRCh38
rs200805334 Chr.11: 107353481-107353481 on Build GRCh38

View Full Product Details