Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33591899-33592156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
UniGene
Hs.468140
Gene Chromosome Location
Chr.2: 33583661-33599362 on Build GRCh38
Transcripts
NM_001304538.1 NM_015475.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3743787 c.320A>G p.N107S Chr.2: 33592097-33592097 on Build GRCh38
5821702 c.284C>A p.S95Y Chr.2: 33592133-33592133 on Build GRCh38
7109816 c.280A>C p.T94P Chr.2: 33592137-33592137 on Build GRCh38
6588397 c.337+8C>G p.? Chr.2: 33592072-33592072 on Build GRCh38
2943194 c.267G>A p.P89P Chr.2: 33592150-33592150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754896581 Chr.2: 33592060-33592060 on Build GRCh38
rs780231907 Chr.2: 33592041-33592041 on Build GRCh38
rs760865035 Chr.2: 33592042-33592043 on Build GRCh38
rs147048996 Chr.2: 33591934-33591934 on Build GRCh38
rs759653803 Chr.2: 33592061-33592061 on Build GRCh38
rs747938716 Chr.2: 33592062-33592062 on Build GRCh38
rs201196734 Chr.2: 33592107-33592107 on Build GRCh38
rs747098345 Chr.2: 33592054-33592054 on Build GRCh38
rs986439457 Chr.2: 33591986-33591986 on Build GRCh38
rs11885608 Chr.2: 33591950-33591950 on Build GRCh38
rs766713089 Chr.2: 33592094-33592094 on Build GRCh38
rs567032920 Chr.2: 33592116-33592116 on Build GRCh38
rs781250542 Chr.2: 33592108-33592108 on Build GRCh38
rs765594708 Chr.2: 33592084-33592084 on Build GRCh38
rs920282335 Chr.2: 33591983-33591983 on Build GRCh38
rs192903923 Chr.2: 33592063-33592063 on Build GRCh38
rs770616395 Chr.2: 33592117-33592117 on Build GRCh38
rs533182270 Chr.2: 33592029-33592029 on Build GRCh38
rs1023443346 Chr.2: 33592122-33592122 on Build GRCh38
rs141905297 Chr.2: 33592071-33592071 on Build GRCh38
rs751681866 Chr.2: 33592096-33592096 on Build GRCh38
rs1015813590 Chr.2: 33591936-33591936 on Build GRCh38
rs750700708 Chr.2: 33592030-33592030 on Build GRCh38
rs758588154 Chr.2: 33592091-33592091 on Build GRCh38
rs762054890 Chr.2: 33592083-33592083 on Build GRCh38
rs764572522 Chr.2: 33592072-33592072 on Build GRCh38
rs375238067 Chr.2: 33592081-33592081 on Build GRCh38
rs143809582 Chr.2: 33592092-33592092 on Build GRCh38
rs1018264059 Chr.2: 33592044-33592044 on Build GRCh38
rs756030331 Chr.2: 33592115-33592115 on Build GRCh38
rs748153796 Chr.2: 33592111-33592111 on Build GRCh38
rs961660848 Chr.2: 33591947-33591947 on Build GRCh38
rs750774826 Chr.2: 33592088-33592088 on Build GRCh38
rs918638129 Chr.2: 33592078-33592078 on Build GRCh38
rs770507226 Chr.2: 33592066-33592066 on Build GRCh38
rs774049008 Chr.2: 33592127-33592127 on Build GRCh38
rs745361069 Chr.2: 33592133-33592133 on Build GRCh38
rs755163482 Chr.2: 33592098-33592098 on Build GRCh38
rs764207387 Chr.2: 33592044-33592044 on Build GRCh38
rs539294537 Chr.2: 33592065-33592065 on Build GRCh38
rs148085074 Chr.2: 33592028-33592028 on Build GRCh38

View Full Product Details