Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35629461-35629732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM42
Entrez Gene ID
79171
Gene Name
RNA binding motif protein 42
Gene Aliases
-
UniGene
Hs.5086
Gene Chromosome Location
Chr.19: 35629016-35637686 on Build GRCh38
Transcripts
NM_001319113.1 NM_024321.4 XM_005259257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6557775 c.137A>T p.Q46L Chr.19: 35629528-35629528 on Build GRCh38
7123496 c.222G>C p.A74A Chr.19: 35629613-35629613 on Build GRCh38
711950 c.202A>G p.T68A Chr.19: 35629593-35629593 on Build GRCh38
6499915 c.222G>A p.A74A Chr.19: 35629613-35629613 on Build GRCh38
4438950 c.164G>A p.G55E Chr.19: 35629555-35629555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762045982 Chr.19: 35629539-35629539 on Build GRCh38
rs774466131 Chr.19: 35629533-35629533 on Build GRCh38
rs747782038 Chr.19: 35629579-35629579 on Build GRCh38
rs776013868 Chr.19: 35629706-35629706 on Build GRCh38
rs527907137 Chr.19: 35629707-35629707 on Build GRCh38
rs762247324 Chr.19: 35629692-35629710 on Build GRCh38
rs775057918 Chr.19: 35629684-35629689 on Build GRCh38
rs560908111 Chr.19: 35629708-35629708 on Build GRCh38
rs769792820 Chr.19: 35629658-35629658 on Build GRCh38
rs1035749593 Chr.19: 35629601-35629601 on Build GRCh38
rs773481616 Chr.19: 35629698-35629698 on Build GRCh38
rs899131065 Chr.19: 35629580-35629580 on Build GRCh38
rs777270224 Chr.19: 35629606-35629606 on Build GRCh38
rs200853423 Chr.19: 35629637-35629637 on Build GRCh38
rs745776620 Chr.19: 35629660-35629660 on Build GRCh38
rs375216406 Chr.19: 35629559-35629559 on Build GRCh38
rs764416047 Chr.19: 35629526-35629526 on Build GRCh38
rs769283488 Chr.19: 35629581-35629589 on Build GRCh38
rs771517189 Chr.19: 35629488-35629488 on Build GRCh38
rs755269298 Chr.19: 35629568-35629568 on Build GRCh38
rs746963812 Chr.19: 35629510-35629510 on Build GRCh38
rs754276246 Chr.19: 35629565-35629565 on Build GRCh38
rs776478765 Chr.19: 35629512-35629512 on Build GRCh38
rs201418448 Chr.19: 35629560-35629560 on Build GRCh38
rs767844889 Chr.19: 35629686-35629686 on Build GRCh38
rs750456872 Chr.19: 35629553-35629553 on Build GRCh38
rs759240557 Chr.19: 35629513-35629513 on Build GRCh38
rs11549651 Chr.19: 35629538-35629538 on Build GRCh38
rs760813931 Chr.19: 35629703-35629703 on Build GRCh38
rs775296610 Chr.19: 35629678-35629678 on Build GRCh38
rs199759784 Chr.19: 35629552-35629552 on Build GRCh38
rs542376420 Chr.19: 35629681-35629681 on Build GRCh38
rs757992768 Chr.19: 35629604-35629604 on Build GRCh38
rs770913769 Chr.19: 35629642-35629642 on Build GRCh38
rs1004731292 Chr.19: 35629632-35629632 on Build GRCh38
rs769497780 Chr.19: 35629675-35629675 on Build GRCh38
rs1034226846 Chr.19: 35629639-35629640 on Build GRCh38
rs765537867 Chr.19: 35629709-35629709 on Build GRCh38
rs776529854 Chr.19: 35629655-35629655 on Build GRCh38
rs779298768 Chr.19: 35629575-35629575 on Build GRCh38
rs761942257 Chr.19: 35629682-35629682 on Build GRCh38

View Full Product Details