Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1717367-1717610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM129
Entrez Gene ID
92305
Gene Name
transmembrane protein 129
Gene Aliases
D4S2561E
UniGene
Hs.518562
Gene Chromosome Location
Chr.4: 1715952-1721357 on Build GRCh38
Transcripts
NM_001127266.1 NM_138385.3 XM_005248038.4 XM_005248039.4 XM_011513594.2 XM_017008834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760478 c.789A>G p.T263T Chr.4: 1717567-1717567 on Build GRCh38
3825535 c.810C>T p.V270V Chr.4: 1717546-1717546 on Build GRCh38
3365553 c.780C>G p.F260L Chr.4: 1717576-1717576 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763646781 Chr.4: 1717513-1717513 on Build GRCh38
rs575169998 Chr.4: 1717584-1717584 on Build GRCh38
rs572935127 Chr.4: 1717541-1717541 on Build GRCh38
rs906365152 Chr.4: 1717445-1717445 on Build GRCh38
rs967552791 Chr.4: 1717538-1717538 on Build GRCh38
rs372844725 Chr.4: 1717448-1717448 on Build GRCh38
rs912167470 Chr.4: 1717524-1717524 on Build GRCh38
rs984879477 Chr.4: 1717525-1717525 on Build GRCh38
rs926130034 Chr.4: 1717554-1717554 on Build GRCh38
rs891933579 Chr.4: 1717433-1717433 on Build GRCh38
rs1006553499 Chr.4: 1717494-1717494 on Build GRCh38
rs760314924 Chr.4: 1717575-1717575 on Build GRCh38
rs767178643 Chr.4: 1717502-1717502 on Build GRCh38
rs936085214 Chr.4: 1717394-1717394 on Build GRCh38
rs928631273 Chr.4: 1717579-1717579 on Build GRCh38
rs746090593 Chr.4: 1717480-1717487 on Build GRCh38
rs1025346334 Chr.4: 1717444-1717444 on Build GRCh38
rs769899482 Chr.4: 1717446-1717446 on Build GRCh38
rs573693181 Chr.4: 1717439-1717439 on Build GRCh38
rs1053713547 Chr.4: 1717418-1717418 on Build GRCh38
rs1042807532 Chr.4: 1717471-1717471 on Build GRCh38
rs536300009 Chr.4: 1717493-1717493 on Build GRCh38
rs889494253 Chr.4: 1717477-1717477 on Build GRCh38
rs376153672 Chr.4: 1717509-1717509 on Build GRCh38
rs770781453 Chr.4: 1717465-1717465 on Build GRCh38
rs544117441 Chr.4: 1717585-1717585 on Build GRCh38
rs781325125 Chr.4: 1717423-1717423 on Build GRCh38
rs552790667 Chr.4: 1717531-1717531 on Build GRCh38
rs762823905 Chr.4: 1717458-1717458 on Build GRCh38
rs374359483 Chr.4: 1717586-1717586 on Build GRCh38
rs980076828 Chr.4: 1717568-1717568 on Build GRCh38
rs1012137266 Chr.4: 1717432-1717432 on Build GRCh38
rs2236786 Chr.4: 1717567-1717567 on Build GRCh38
rs1055782415 Chr.4: 1717395-1717395 on Build GRCh38
rs768916395 Chr.4: 1717422-1717422 on Build GRCh38
rs112780538 Chr.4: 1717488-1717488 on Build GRCh38
rs774193075 Chr.4: 1717472-1717472 on Build GRCh38
rs566573197 Chr.4: 1717504-1717504 on Build GRCh38
rs544861212 Chr.4: 1717561-1717561 on Build GRCh38
rs747338831 Chr.4: 1717387-1717387 on Build GRCh38

View Full Product Details