Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71376968-71377221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF1
Entrez Gene ID
6872
Gene Name
TATA-box binding protein associated factor 1
Gene Aliases
BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP
UniGene
Hs.158560
Gene Chromosome Location
Chr.X: 71366220-71530525 on Build GRCh38
Transcripts
NM_001286074.1 NM_004606.4 NM_138923.3 NR_104387.1 NR_104388.1 NR_104389.1 NR_104390.1 NR_104391.1 NR_104392.1 NR_104393.1 NR_104394.1 NR_104395.1 XM_005262295.1 XM_005262296.1 XM_005262297.4 XM_005262300.2 XM_006724682.3

SNP Detail

SNP ID Chromosome Location
rs753944450 Chr.X: 71377143-71377143 on Build GRCh38
rs1002075475 Chr.X: 71377127-71377127 on Build GRCh38
rs538436943 Chr.X: 71377140-71377140 on Build GRCh38
rs1021042308 Chr.X: 71377147-71377147 on Build GRCh38
rs778363507 Chr.X: 71377196-71377196 on Build GRCh38
rs764474845 Chr.X: 71376992-71376992 on Build GRCh38
rs866584662 Chr.X: 71377109-71377109 on Build GRCh38
rs753114489 Chr.X: 71377161-71377161 on Build GRCh38
rs772975167 Chr.X: 71377061-71377061 on Build GRCh38
rs1051551145 Chr.X: 71377027-71377027 on Build GRCh38
rs758629598 Chr.X: 71377194-71377194 on Build GRCh38
rs773107206 Chr.X: 71377110-71377110 on Build GRCh38
rs755308739 Chr.X: 71377154-71377154 on Build GRCh38
rs151257934 Chr.X: 71377160-71377160 on Build GRCh38
rs747430823 Chr.X: 71377199-71377199 on Build GRCh38
rs891180704 Chr.X: 71377088-71377088 on Build GRCh38
rs571774489 Chr.X: 71377130-71377130 on Build GRCh38
rs767505496 Chr.X: 71377004-71377004 on Build GRCh38

View Full Product Details