Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 46776779-46777026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A4
Entrez Gene ID
55089
Gene Name
solute carrier family 38 member 4
Gene Aliases
ATA3, NAT3, PAAT, SNAT4
UniGene
Hs.446077
Gene Chromosome Location
Chr.12: 46764761-46832422 on Build GRCh38
Transcripts
NM_001143824.1 NM_018018.4 XM_005268997.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4727290 c.1126C>T p.L376F Chr.12: 46776952-46776952 on Build GRCh38
5435270 c.1097C>T p.T366M Chr.12: 46776981-46776981 on Build GRCh38
72692 c.1145C>T p.A382V Chr.12: 46776933-46776933 on Build GRCh38
5375198 c.1085G>A p.R362K Chr.12: 46776993-46776993 on Build GRCh38
6782009 c.1156G>T p.G386C Chr.12: 46776922-46776922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147575073 Chr.12: 46776960-46776960 on Build GRCh38
rs756151783 Chr.12: 46776959-46776959 on Build GRCh38
rs777458182 Chr.12: 46776892-46776892 on Build GRCh38
rs576025485 Chr.12: 46776826-46776826 on Build GRCh38
rs373742937 Chr.12: 46776931-46776931 on Build GRCh38
rs776123077 Chr.12: 46776923-46776923 on Build GRCh38
rs145123611 Chr.12: 46776926-46776926 on Build GRCh38
rs559154095 Chr.12: 46776817-46776817 on Build GRCh38
rs768636628 Chr.12: 46776996-46776996 on Build GRCh38
rs754857011 Chr.12: 46776874-46776874 on Build GRCh38
rs573543607 Chr.12: 46776845-46776845 on Build GRCh38
rs150492267 Chr.12: 46776997-46776997 on Build GRCh38
rs770362299 Chr.12: 46776914-46776914 on Build GRCh38
rs776159143 Chr.12: 46776919-46776919 on Build GRCh38
rs910998311 Chr.12: 46776965-46776965 on Build GRCh38
rs762033982 Chr.12: 46776948-46776948 on Build GRCh38
rs201247215 Chr.12: 46776980-46776980 on Build GRCh38
rs746667603 Chr.12: 46776905-46776905 on Build GRCh38
rs542750618 Chr.12: 46776879-46776879 on Build GRCh38
rs751708832 Chr.12: 46776993-46776993 on Build GRCh38
rs370296366 Chr.12: 46776856-46776856 on Build GRCh38
rs12316767 Chr.12: 46776838-46776838 on Build GRCh38
rs371647665 Chr.12: 46776812-46776812 on Build GRCh38
rs957345596 Chr.12: 46776875-46776875 on Build GRCh38
rs750472336 Chr.12: 46776950-46776950 on Build GRCh38
rs771831340 Chr.12: 46776887-46776887 on Build GRCh38
rs747976495 Chr.12: 46776882-46776882 on Build GRCh38
rs770465170 Chr.12: 46776921-46776921 on Build GRCh38
rs985628064 Chr.12: 46776955-46776955 on Build GRCh38
rs550174304 Chr.12: 46776949-46776949 on Build GRCh38
rs11183610 Chr.12: 46776981-46776981 on Build GRCh38
rs966653230 Chr.12: 46776898-46776898 on Build GRCh38
rs375605342 Chr.12: 46776932-46776932 on Build GRCh38
rs77993723 Chr.12: 46776834-46776834 on Build GRCh38
rs1011607757 Chr.12: 46776877-46776877 on Build GRCh38

View Full Product Details