Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138917510-138917737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REPS1
Entrez Gene ID
85021
Gene Name
RALBP1 associated Eps domain containing 1
Gene Aliases
RALBP1
UniGene
Hs.334603
Gene Chromosome Location
Chr.6: 138904015-138988282 on Build GRCh38
Transcripts
NM_001128617.2 NM_001286611.1 NM_001286612.1 NM_031922.4 XM_005267177.3 XM_005267178.4 XM_005267179.3 XM_011536202.2 XM_017011387.1 XM_017011388.1 XM_017011389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5171292 c.1397_1398delTT p.F466fs*3 Chr.6: 138917599-138917600 on Build GRCh38
6309930 c.1442+7A>G p.? Chr.6: 138917548-138917548 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752858373 Chr.6: 138917607-138917607 on Build GRCh38
rs756188622 Chr.6: 138917615-138917615 on Build GRCh38
rs1035512626 Chr.6: 138917711-138917711 on Build GRCh38
rs773955524 Chr.6: 138917661-138917665 on Build GRCh38
rs140498560 Chr.6: 138917675-138917675 on Build GRCh38
rs755325656 Chr.6: 138917582-138917582 on Build GRCh38
rs761479306 Chr.6: 138917670-138917671 on Build GRCh38
rs767747902 Chr.6: 138917589-138917589 on Build GRCh38
rs771654246 Chr.6: 138917561-138917561 on Build GRCh38
rs774064933 Chr.6: 138917540-138917540 on Build GRCh38
rs751904980 Chr.6: 138917578-138917578 on Build GRCh38
rs981807284 Chr.6: 138917700-138917700 on Build GRCh38
rs777894001 Chr.6: 138917636-138917636 on Build GRCh38
rs572381894 Chr.6: 138917690-138917690 on Build GRCh38
rs368745071 Chr.6: 138917538-138917538 on Build GRCh38
rs763635845 Chr.6: 138917629-138917630 on Build GRCh38
rs749367138 Chr.6: 138917637-138917637 on Build GRCh38
rs766884413 Chr.6: 138917576-138917576 on Build GRCh38
rs553814177 Chr.6: 138917570-138917570 on Build GRCh38
rs759344632 Chr.6: 138917558-138917558 on Build GRCh38
rs1045779635 Chr.6: 138917590-138917590 on Build GRCh38
rs775237818 Chr.6: 138917569-138917569 on Build GRCh38
rs994109110 Chr.6: 138917685-138917685 on Build GRCh38

View Full Product Details