Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177933467-177933692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC16B
Entrez Gene ID
89866
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Gene Aliases
LZTR2, PGPR-p117, RGPR, SEC16S
UniGene
Hs.149540
Gene Chromosome Location
Chr.1: 177929107-177969915 on Build GRCh38
Transcripts
NM_033127.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6774106 c.2675G>A p.R892Q Chr.1: 177933533-177933533 on Build GRCh38
4981754 c.2621C>T p.S874L Chr.1: 177933587-177933587 on Build GRCh38
4608243 c.2621C>A p.S874* Chr.1: 177933587-177933587 on Build GRCh38
5446669 c.2582C>T p.A861V Chr.1: 177933626-177933626 on Build GRCh38
3997084 c.2618G>A p.S873N Chr.1: 177933590-177933590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369485878 Chr.1: 177933594-177933594 on Build GRCh38
rs1028969296 Chr.1: 177933584-177933584 on Build GRCh38
rs904797281 Chr.1: 177933585-177933585 on Build GRCh38
rs543330759 Chr.1: 177933614-177933614 on Build GRCh38
rs760101140 Chr.1: 177933495-177933495 on Build GRCh38
rs540830662 Chr.1: 177933543-177933543 on Build GRCh38
rs887155080 Chr.1: 177933493-177933493 on Build GRCh38
rs751704250 Chr.1: 177933599-177933599 on Build GRCh38
rs770450983 Chr.1: 177933540-177933540 on Build GRCh38
rs3813649 Chr.1: 177933590-177933590 on Build GRCh38
rs183052044 Chr.1: 177933576-177933576 on Build GRCh38
rs591120 Chr.1: 177933618-177933618 on Build GRCh38
rs769133118 Chr.1: 177933549-177933549 on Build GRCh38
rs776148451 Chr.1: 177933581-177933581 on Build GRCh38
rs770317188 Chr.1: 177933574-177933574 on Build GRCh38
rs373528898 Chr.1: 177933595-177933595 on Build GRCh38
rs770638639 Chr.1: 177933634-177933634 on Build GRCh38
rs192490446 Chr.1: 177933639-177933639 on Build GRCh38
rs754949803 Chr.1: 177933507-177933507 on Build GRCh38
rs757536439 Chr.1: 177933600-177933600 on Build GRCh38
rs578014026 Chr.1: 177933494-177933494 on Build GRCh38
rs369332659 Chr.1: 177933508-177933508 on Build GRCh38
rs557959940 Chr.1: 177933491-177933491 on Build GRCh38
rs781377407 Chr.1: 177933538-177933538 on Build GRCh38
rs753697048 Chr.1: 177933500-177933500 on Build GRCh38
rs187677582 Chr.1: 177933620-177933620 on Build GRCh38
rs1048268784 Chr.1: 177933490-177933490 on Build GRCh38
rs759419318 Chr.1: 177933582-177933582 on Build GRCh38
rs375305713 Chr.1: 177933496-177933496 on Build GRCh38
rs773732478 Chr.1: 177933568-177933570 on Build GRCh38
rs772728174 Chr.1: 177933552-177933552 on Build GRCh38
rs370816982 Chr.1: 177933562-177933562 on Build GRCh38
rs777861102 Chr.1: 177933533-177933533 on Build GRCh38
rs760425203 Chr.1: 177933554-177933554 on Build GRCh38
rs776062221 Chr.1: 177933636-177933636 on Build GRCh38
rs756624716 Chr.1: 177933610-177933610 on Build GRCh38
rs749677926 Chr.1: 177933546-177933546 on Build GRCh38
rs758555753 Chr.1: 177933529-177933529 on Build GRCh38
rs746603191 Chr.1: 177933630-177933630 on Build GRCh38
rs746342263 Chr.1: 177933539-177933539 on Build GRCh38
rs891949546 Chr.1: 177933580-177933580 on Build GRCh38
rs377454481 Chr.1: 177933605-177933605 on Build GRCh38
rs377409580 Chr.1: 177933524-177933524 on Build GRCh38
rs901203307 Chr.1: 177933534-177933534 on Build GRCh38
rs371931365 Chr.1: 177933537-177933537 on Build GRCh38

View Full Product Details