Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69653243-69653494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFAT5
Entrez Gene ID
10725
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
Gene Aliases
NF-AT5, NFATL1, NFATZ, OREBP, TONEBP
UniGene
Hs.371987
Gene Chromosome Location
Chr.16: 69565952-69704666 on Build GRCh38
Transcripts
NM_001113178.2 NM_006599.3 NM_138713.3 NM_138714.3 NM_173214.2 NM_173215.2 XM_006721125.3 XM_011522817.2 XM_011522818.2 XM_011522819.2 XM_011522820.2 XM_011522822.2 XM_017022870.1 XM_017022871.1 XM_017022872.1 XM_017022873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015392 c.965C>G p.S322* Chr.16: 69653388-69653388 on Build GRCh38
4062246 c.841G>A p.V281I Chr.16: 69653264-69653264 on Build GRCh38
224505 c.602G>A p.S201N Chr.16: 69653307-69653307 on Build GRCh38
4062247 c.559G>A p.V187I Chr.16: 69653264-69653264 on Build GRCh38
6725122 c.934G>A p.A312T Chr.16: 69653357-69653357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779894815 Chr.16: 69653362-69653362 on Build GRCh38
rs761214173 Chr.16: 69653446-69653446 on Build GRCh38
rs759840029 Chr.16: 69653454-69653454 on Build GRCh38
rs774078368 Chr.16: 69653398-69653398 on Build GRCh38
rs12923676 Chr.16: 69653372-69653372 on Build GRCh38
rs751067427 Chr.16: 69653437-69653440 on Build GRCh38
rs773641818 Chr.16: 69653435-69653435 on Build GRCh38
rs766724495 Chr.16: 69653447-69653447 on Build GRCh38
rs201220463 Chr.16: 69653306-69653306 on Build GRCh38
rs1036781757 Chr.16: 69653275-69653275 on Build GRCh38
rs748951572 Chr.16: 69653377-69653377 on Build GRCh38
rs147145018 Chr.16: 69653287-69653287 on Build GRCh38
rs777044916 Chr.16: 69653451-69653451 on Build GRCh38
rs867909989 Chr.16: 69653354-69653354 on Build GRCh38
rs1017724743 Chr.16: 69653429-69653430 on Build GRCh38
rs765362536 Chr.16: 69653286-69653286 on Build GRCh38
rs201684496 Chr.16: 69653432-69653432 on Build GRCh38
rs530092980 Chr.16: 69653293-69653293 on Build GRCh38
rs768435664 Chr.16: 69653381-69653381 on Build GRCh38
rs865774873 Chr.16: 69653353-69653353 on Build GRCh38
rs755898607 Chr.16: 69653300-69653300 on Build GRCh38
rs747694582 Chr.16: 69653422-69653422 on Build GRCh38
rs779266189 Chr.16: 69653451-69653465 on Build GRCh38
rs781126963 Chr.16: 69653292-69653292 on Build GRCh38
rs758430767 Chr.16: 69653290-69653290 on Build GRCh38

View Full Product Details