Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30007081-30007354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6818
Entrez Gene ID
102466742
Gene Name
microRNA 6818
Gene Aliases
hsa-mir-6818
UniGene
Gene Chromosome Location
Chr.22: 30007049-30007113 on Build GRCh38
Transcripts
NR_106876.1 NM_021090.3 NM_153050.2 NM_153051.2 XM_005261803.2 XM_005261804.3 XM_005261805.2 XM_005261806.2 XM_005261808.2 XM_017029028.1 XM_017029029.1 XM_017029030.1 XM_017029031.1 XM_017029032.1 XM_017029033.1 XM_017029034.1

Gene Symbol
MTMR3
Entrez Gene ID
8897
Gene Name
myotubularin related protein 3
Gene Aliases
FYVE-DSP1, ZFYVE10
UniGene
Hs.474536
Gene Chromosome Location
Chr.22: 29883165-30030868 on Build GRCh38
Transcripts
NR_106876.1 NM_021090.3 NM_153050.2 NM_153051.2 XM_005261803.2 XM_005261804.3 XM_005261805.2 XM_005261806.2 XM_005261808.2 XM_017029028.1 XM_017029029.1 XM_017029030.1 XM_017029031.1 XM_017029032.1 XM_017029033.1 XM_017029034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103381 c.701G>A p.R234H Chr.22: 30007143-30007143 on Build GRCh38
1291236 c.874T>G p.F292V Chr.22: 30007316-30007316 on Build GRCh38
4771365 c.759G>T p.L253L Chr.22: 30007201-30007201 on Build GRCh38
4771364 c.759G>T p.L253L Chr.22: 30007201-30007201 on Build GRCh38
7091286 c.848A>G p.N283S Chr.22: 30007290-30007290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774582819 Chr.22: 30007136-30007136 on Build GRCh38
rs770933269 Chr.22: 30007239-30007239 on Build GRCh38
rs145026900 Chr.22: 30007233-30007233 on Build GRCh38
rs999063533 Chr.22: 30007310-30007310 on Build GRCh38
rs947920440 Chr.22: 30007243-30007243 on Build GRCh38
rs138787389 Chr.22: 30007290-30007290 on Build GRCh38
rs756629957 Chr.22: 30007285-30007285 on Build GRCh38
rs778391690 Chr.22: 30007160-30007160 on Build GRCh38
rs771687621 Chr.22: 30007210-30007210 on Build GRCh38
rs146523702 Chr.22: 30007318-30007318 on Build GRCh38
rs750960176 Chr.22: 30007310-30007312 on Build GRCh38
rs776057188 Chr.22: 30007253-30007253 on Build GRCh38
rs918610010 Chr.22: 30007125-30007125 on Build GRCh38
rs976429195 Chr.22: 30007117-30007117 on Build GRCh38
rs759208228 Chr.22: 30007246-30007246 on Build GRCh38
rs761917081 Chr.22: 30007274-30007274 on Build GRCh38
rs930048625 Chr.22: 30007126-30007126 on Build GRCh38
rs892137207 Chr.22: 30007309-30007309 on Build GRCh38
rs755250944 Chr.22: 30007293-30007293 on Build GRCh38
rs537954673 Chr.22: 30007242-30007242 on Build GRCh38
rs764395988 Chr.22: 30007252-30007252 on Build GRCh38
rs202174195 Chr.22: 30007301-30007301 on Build GRCh38
rs724159917 Chr.22: 30007277-30007277 on Build GRCh38
rs766732507 Chr.22: 30007286-30007286 on Build GRCh38
rs774410241 Chr.22: 30007270-30007270 on Build GRCh38
rs780455916 Chr.22: 30007122-30007122 on Build GRCh38
rs892905064 Chr.22: 30007331-30007331 on Build GRCh38
rs778676274 Chr.22: 30007294-30007294 on Build GRCh38
rs567791660 Chr.22: 30007159-30007159 on Build GRCh38
rs377428278 Chr.22: 30007204-30007204 on Build GRCh38
rs750990316 Chr.22: 30007280-30007280 on Build GRCh38
rs773348982 Chr.22: 30007228-30007228 on Build GRCh38
rs868055178 Chr.22: 30007208-30007208 on Build GRCh38
rs767584169 Chr.22: 30007278-30007278 on Build GRCh38

View Full Product Details