Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111708146-111708419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCXD2
Entrez Gene ID
257068
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Gene Aliases
-
UniGene
Hs.477114
Gene Chromosome Location
Chr.3: 111674676-111846447 on Build GRCh38
Transcripts
NM_001185106.1 NM_153268.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585221 c.393C>T p.I131I Chr.3: 111708155-111708155 on Build GRCh38
6748474 c.401delT p.F135fs*9 Chr.3: 111708163-111708163 on Build GRCh38
4317674 c.411C>G p.I137M Chr.3: 111708173-111708173 on Build GRCh38
4583784 c.535C>T p.R179W Chr.3: 111708297-111708297 on Build GRCh38
727294 c.389T>C p.F130S Chr.3: 111708151-111708151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778426481 Chr.3: 111708259-111708259 on Build GRCh38
rs754576362 Chr.3: 111708240-111708240 on Build GRCh38
rs896253181 Chr.3: 111708272-111708272 on Build GRCh38
rs917559079 Chr.3: 111708292-111708292 on Build GRCh38
rs371366632 Chr.3: 111708277-111708277 on Build GRCh38
rs748756039 Chr.3: 111708364-111708365 on Build GRCh38
rs746786741 Chr.3: 111708358-111708358 on Build GRCh38
rs745756251 Chr.3: 111708364-111708364 on Build GRCh38
rs751872957 Chr.3: 111708206-111708206 on Build GRCh38
rs778430646 Chr.3: 111708342-111708342 on Build GRCh38
rs200714034 Chr.3: 111708204-111708204 on Build GRCh38
rs770776640 Chr.3: 111708360-111708360 on Build GRCh38
rs574958079 Chr.3: 111708297-111708297 on Build GRCh38
rs774448388 Chr.3: 111708385-111708385 on Build GRCh38
rs777498961 Chr.3: 111708274-111708274 on Build GRCh38
rs766977855 Chr.3: 111708337-111708337 on Build GRCh38
rs183121832 Chr.3: 111708395-111708395 on Build GRCh38
rs771725194 Chr.3: 111708268-111708268 on Build GRCh38
rs762042092 Chr.3: 111708310-111708310 on Build GRCh38
rs1047909737 Chr.3: 111708345-111708345 on Build GRCh38
rs757421234 Chr.3: 111708221-111708221 on Build GRCh38
rs775510663 Chr.3: 111708175-111708175 on Build GRCh38
rs144525978 Chr.3: 111708201-111708201 on Build GRCh38
rs201083256 Chr.3: 111708223-111708223 on Build GRCh38
rs762024556 Chr.3: 111708388-111708388 on Build GRCh38
rs914687559 Chr.3: 111708186-111708186 on Build GRCh38
rs752170813 Chr.3: 111708349-111708349 on Build GRCh38
rs367817783 Chr.3: 111708298-111708298 on Build GRCh38
rs768712026 Chr.3: 111708304-111708304 on Build GRCh38
rs754437797 Chr.3: 111708338-111708338 on Build GRCh38
rs756648742 Chr.3: 111708236-111708236 on Build GRCh38
rs770592107 Chr.3: 111708373-111708375 on Build GRCh38
rs750839141 Chr.3: 111708228-111708228 on Build GRCh38
rs748722940 Chr.3: 111708239-111708239 on Build GRCh38
rs769637051 Chr.3: 111708384-111708384 on Build GRCh38
rs746903071 Chr.3: 111708329-111708329 on Build GRCh38
rs1056195742 Chr.3: 111708271-111708271 on Build GRCh38
rs937935486 Chr.3: 111708262-111708262 on Build GRCh38
rs201796337 Chr.3: 111708361-111708361 on Build GRCh38
rs967833823 Chr.3: 111708202-111708202 on Build GRCh38
rs770445265 Chr.3: 111708355-111708355 on Build GRCh38
rs887538682 Chr.3: 111708353-111708353 on Build GRCh38
rs370255847 Chr.3: 111708325-111708325 on Build GRCh38
rs762935728 Chr.3: 111708299-111708299 on Build GRCh38
rs556720268 Chr.3: 111708390-111708390 on Build GRCh38
rs774304260 Chr.3: 111708309-111708309 on Build GRCh38
rs777445571 Chr.3: 111708356-111708356 on Build GRCh38
rs373820996 Chr.3: 111708280-111708280 on Build GRCh38
rs199840687 Chr.3: 111708341-111708341 on Build GRCh38
rs761170107 Chr.3: 111708327-111708327 on Build GRCh38
rs780745161 Chr.3: 111708238-111708238 on Build GRCh38

View Full Product Details