Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168811490-168811731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOSTRIN
Entrez Gene ID
115677
Gene Name
nitric oxide synthase trafficking
Gene Aliases
DaIP2
UniGene
Hs.189780
Gene Chromosome Location
Chr.2: 168786539-168865339 on Build GRCh38
Transcripts
NM_001039724.3 NM_001171631.1 NM_001171632.1 NM_052946.3 XM_005246270.1 XM_006712233.3 XM_011510540.2 XM_017003278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1259317 c.62C>G p.S21C Chr.2: 168811601-168811601 on Build GRCh38
5782480 c.73G>C p.E25Q Chr.2: 168811612-168811612 on Build GRCh38
6192472 c.45G>A p.K15K Chr.2: 168811584-168811584 on Build GRCh38
1009076 c.75G>T p.E25D Chr.2: 168811614-168811614 on Build GRCh38
5782481 c.73G>C p.E25Q Chr.2: 168811612-168811612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915681179 Chr.2: 168811535-168811535 on Build GRCh38
rs748722343 Chr.2: 168811637-168811637 on Build GRCh38
rs781506508 Chr.2: 168811627-168811627 on Build GRCh38
rs371981545 Chr.2: 168811679-168811679 on Build GRCh38
rs763150815 Chr.2: 168811539-168811539 on Build GRCh38
rs766551545 Chr.2: 168811678-168811678 on Build GRCh38
rs772705309 Chr.2: 168811519-168811519 on Build GRCh38
rs368106312 Chr.2: 168811579-168811579 on Build GRCh38
rs766659292 Chr.2: 168811610-168811610 on Build GRCh38
rs368583308 Chr.2: 168811520-168811520 on Build GRCh38
rs529424492 Chr.2: 168811605-168811605 on Build GRCh38
rs770306756 Chr.2: 168811663-168811663 on Build GRCh38
rs764135471 Chr.2: 168811680-168811680 on Build GRCh38
rs767164672 Chr.2: 168811553-168811553 on Build GRCh38
rs80075099 Chr.2: 168811612-168811612 on Build GRCh38
rs551557124 Chr.2: 168811538-168811539 on Build GRCh38
rs960021821 Chr.2: 168811529-168811529 on Build GRCh38
rs756601575 Chr.2: 168811639-168811639 on Build GRCh38
rs778192785 Chr.2: 168811648-168811648 on Build GRCh38
rs976999849 Chr.2: 168811530-168811530 on Build GRCh38
rs763618110 Chr.2: 168811618-168811618 on Build GRCh38
rs760491214 Chr.2: 168811670-168811670 on Build GRCh38
rs942259229 Chr.2: 168811686-168811686 on Build GRCh38
rs771170222 Chr.2: 168811556-168811557 on Build GRCh38
rs759406033 Chr.2: 168811546-168811546 on Build GRCh38
rs923319637 Chr.2: 168811532-168811532 on Build GRCh38
rs773807505 Chr.2: 168811541-168811541 on Build GRCh38
rs191918822 Chr.2: 168811665-168811665 on Build GRCh38
rs775417955 Chr.2: 168811667-168811667 on Build GRCh38
rs758345758 Chr.2: 168811593-168811593 on Build GRCh38
rs775614192 Chr.2: 168811574-168811574 on Build GRCh38
rs770628575 Chr.2: 168811531-168811531 on Build GRCh38
rs142731354 Chr.2: 168811583-168811583 on Build GRCh38
rs925515991 Chr.2: 168811677-168811677 on Build GRCh38
rs765477107 Chr.2: 168811581-168811581 on Build GRCh38
rs755419665 Chr.2: 168811623-168811623 on Build GRCh38
rs748799918 Chr.2: 168811657-168811657 on Build GRCh38

View Full Product Details