Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147881413-147881658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf46
Entrez Gene ID
389336
Gene Name
chromosome 5 open reading frame 46
Gene Aliases
SSSP1
UniGene
Hs.660038
Gene Chromosome Location
Chr.5: 147850463-147906594 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Gene Symbol
SCGB3A2
Entrez Gene ID
117156
Gene Name
secretoglobin family 3A member 2
Gene Aliases
LU103, PNSP1, UGRP1, pnSP-1
UniGene
Hs.483765
Gene Chromosome Location
Chr.5: 147878711-147882193 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3852526 c.181G>C p.E61Q Chr.5: 147881571-147881571 on Build GRCh38
1063952 c.73A>G p.N25D Chr.5: 147881463-147881463 on Build GRCh38
4141545 c.255G>A p.L85L Chr.5: 147881645-147881645 on Build GRCh38
3768228 c.254T>C p.L85P Chr.5: 147881644-147881644 on Build GRCh38
6772683 c.82C>T p.P28S Chr.5: 147881472-147881472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750911275 Chr.5: 147881562-147881562 on Build GRCh38
rs745415240 Chr.5: 147881541-147881541 on Build GRCh38
rs746750493 Chr.5: 147881472-147881472 on Build GRCh38
rs758462858 Chr.5: 147881446-147881446 on Build GRCh38
rs750914244 Chr.5: 147881445-147881445 on Build GRCh38
rs140599638 Chr.5: 147881560-147881560 on Build GRCh38
rs746454732 Chr.5: 147881635-147881635 on Build GRCh38
rs770723371 Chr.5: 147881493-147881493 on Build GRCh38
rs760362085 Chr.5: 147881520-147881520 on Build GRCh38
rs755052157 Chr.5: 147881586-147881586 on Build GRCh38
rs774916078 Chr.5: 147881517-147881517 on Build GRCh38
rs771962468 Chr.5: 147881617-147881617 on Build GRCh38
rs1004343272 Chr.5: 147881460-147881460 on Build GRCh38
rs765774292 Chr.5: 147881441-147881441 on Build GRCh38
rs140212139 Chr.5: 147881597-147881597 on Build GRCh38
rs763418998 Chr.5: 147881571-147881571 on Build GRCh38
rs757143528 Chr.5: 147881541-147881546 on Build GRCh38
rs922958334 Chr.5: 147881499-147881499 on Build GRCh38
rs556241547 Chr.5: 147881481-147881481 on Build GRCh38
rs140936862 Chr.5: 147881463-147881463 on Build GRCh38
rs745826855 Chr.5: 147881513-147881513 on Build GRCh38
rs747232043 Chr.5: 147881464-147881464 on Build GRCh38
rs755248204 Chr.5: 147881472-147881472 on Build GRCh38
rs150100560 Chr.5: 147881582-147881582 on Build GRCh38
rs745799049 Chr.5: 147881602-147881602 on Build GRCh38
rs763908581 Chr.5: 147881539-147881539 on Build GRCh38
rs778616621 Chr.5: 147881601-147881601 on Build GRCh38
rs779612264 Chr.5: 147881625-147881625 on Build GRCh38
rs146666696 Chr.5: 147881585-147881585 on Build GRCh38
rs200621359 Chr.5: 147881457-147881457 on Build GRCh38
rs142388312 Chr.5: 147881485-147881485 on Build GRCh38
rs757007939 Chr.5: 147881599-147881599 on Build GRCh38
rs774298931 Chr.5: 147881496-147881496 on Build GRCh38
rs151333009 Chr.5: 147881550-147881550 on Build GRCh38
rs780518946 Chr.5: 147881540-147881540 on Build GRCh38
rs961784042 Chr.5: 147881473-147881473 on Build GRCh38
rs771584795 Chr.5: 147881514-147881514 on Build GRCh38
rs374818057 Chr.5: 147881437-147881437 on Build GRCh38

View Full Product Details