Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22034393-22034666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPM2
Entrez Gene ID
10361
Gene Name
nucleophosmin/nucleoplasmin 2
Gene Aliases
-
UniGene
Hs.131055
Gene Chromosome Location
Chr.8: 22023308-22036908 on Build GRCh38
Transcripts
NM_001286680.1 NM_001286681.1 NM_182795.1 XM_011544360.2 XM_011544362.2 XM_011544363.2 XM_011544364.2 XM_017012947.1 XM_017012948.1 XM_017012949.1 XM_017012950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6472477 c.532-9G>T p.? Chr.8: 22034501-22034501 on Build GRCh38
4707840 c.532delA p.K180fs*9 Chr.8: 22034510-22034510 on Build GRCh38
5119983 c.566+7C>A p.? Chr.8: 22034551-22034551 on Build GRCh38
6472478 c.533A>G p.K178R Chr.8: 22034511-22034511 on Build GRCh38
7111938 c.532A>G p.K178E Chr.8: 22034510-22034510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996475027 Chr.8: 22034616-22034616 on Build GRCh38
rs10089142 Chr.8: 22034428-22034428 on Build GRCh38
rs748573441 Chr.8: 22034477-22034477 on Build GRCh38
rs748826494 Chr.8: 22034647-22034647 on Build GRCh38
rs757125644 Chr.8: 22034533-22034533 on Build GRCh38
rs770395794 Chr.8: 22034526-22034528 on Build GRCh38
rs778184465 Chr.8: 22034598-22034598 on Build GRCh38
rs1028115111 Chr.8: 22034620-22034620 on Build GRCh38
rs747210964 Chr.8: 22034552-22034552 on Build GRCh38
rs775856522 Chr.8: 22034430-22034430 on Build GRCh38
rs765161221 Chr.8: 22034536-22034536 on Build GRCh38
rs117230258 Chr.8: 22034542-22034542 on Build GRCh38
rs192854274 Chr.8: 22034503-22034503 on Build GRCh38
rs775991930 Chr.8: 22034510-22034510 on Build GRCh38
rs773644836 Chr.8: 22034583-22034583 on Build GRCh38
rs373299101 Chr.8: 22034427-22034427 on Build GRCh38
rs917935839 Chr.8: 22034433-22034433 on Build GRCh38
rs770218874 Chr.8: 22034574-22034574 on Build GRCh38
rs4871915 Chr.8: 22034457-22034457 on Build GRCh38
rs373502672 Chr.8: 22034596-22034596 on Build GRCh38
rs1019846940 Chr.8: 22034593-22034593 on Build GRCh38
rs1008364628 Chr.8: 22034414-22034414 on Build GRCh38
rs780444742 Chr.8: 22034544-22034544 on Build GRCh38
rs746576416 Chr.8: 22034494-22034495 on Build GRCh38
rs564976235 Chr.8: 22034468-22034468 on Build GRCh38
rs777243407 Chr.8: 22034471-22034471 on Build GRCh38
rs758991699 Chr.8: 22034490-22034490 on Build GRCh38
rs200348100 Chr.8: 22034529-22034529 on Build GRCh38
rs780354790 Chr.8: 22034462-22034462 on Build GRCh38
rs746427922 Chr.8: 22034522-22034527 on Build GRCh38
rs376506214 Chr.8: 22034594-22034594 on Build GRCh38
rs755411158 Chr.8: 22034556-22034556 on Build GRCh38
rs777493838 Chr.8: 22034471-22034474 on Build GRCh38
rs988678042 Chr.8: 22034597-22034597 on Build GRCh38
rs767163549 Chr.8: 22034500-22034500 on Build GRCh38
rs773839137 Chr.8: 22034487-22034487 on Build GRCh38
rs1045136854 Chr.8: 22034458-22034458 on Build GRCh38
rs770542504 Chr.8: 22034481-22034481 on Build GRCh38
rs753876663 Chr.8: 22034531-22034531 on Build GRCh38
rs747407172 Chr.8: 22034467-22034467 on Build GRCh38
rs957310380 Chr.8: 22034558-22034558 on Build GRCh38
rs760489935 Chr.8: 22034517-22034517 on Build GRCh38
rs750532537 Chr.8: 22034537-22034537 on Build GRCh38
rs949942688 Chr.8: 22034629-22034629 on Build GRCh38
rs748748509 Chr.8: 22034560-22034560 on Build GRCh38
rs770540772 Chr.8: 22034509-22034510 on Build GRCh38
rs1045529373 Chr.8: 22034630-22034630 on Build GRCh38
rs781406892 Chr.8: 22034557-22034557 on Build GRCh38

View Full Product Details