Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43191263-43191520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBR1
Entrez Gene ID
4077
Gene Name
NBR1, autophagy cargo receptor
Gene Aliases
1A1-3B, IAI3B, M17S2, MIG19
UniGene
Hs.277721
Gene Chromosome Location
Chr.17: 43170471-43211689 on Build GRCh38
Transcripts
NM_001291571.1 NM_001291572.1 NM_005899.4 NM_031862.3 XM_006721903.1 XM_011524813.1 XM_017024642.1 XM_017024643.1 XM_017024644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7379011 c.880G>C p.D294H Chr.17: 43191388-43191388 on Build GRCh38
3517902 c.967C>T p.L323F Chr.17: 43191475-43191475 on Build GRCh38
4414010 c.914T>C p.L305S Chr.17: 43191422-43191422 on Build GRCh38
1258768 c.974T>C p.L325P Chr.17: 43191482-43191482 on Build GRCh38
5755049 c.877G>A p.V293I Chr.17: 43191385-43191385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561234403 Chr.17: 43191300-43191300 on Build GRCh38
rs778277020 Chr.17: 43191455-43191455 on Build GRCh38
rs1017480816 Chr.17: 43191448-43191448 on Build GRCh38
rs147136918 Chr.17: 43191294-43191294 on Build GRCh38
rs756071973 Chr.17: 43191423-43191423 on Build GRCh38
rs994697255 Chr.17: 43191480-43191480 on Build GRCh38
rs148639609 Chr.17: 43191314-43191314 on Build GRCh38
rs774547644 Chr.17: 43191400-43191400 on Build GRCh38
rs763591262 Chr.17: 43191385-43191385 on Build GRCh38
rs967316797 Chr.17: 43191396-43191396 on Build GRCh38
rs773632845 Chr.17: 43191375-43191375 on Build GRCh38
rs532094208 Chr.17: 43191484-43191484 on Build GRCh38
rs750178073 Chr.17: 43191429-43191429 on Build GRCh38
rs761883172 Chr.17: 43191424-43191424 on Build GRCh38
rs892778572 Chr.17: 43191316-43191316 on Build GRCh38
rs755927037 Chr.17: 43191442-43191442 on Build GRCh38
rs746241930 Chr.17: 43191346-43191346 on Build GRCh38
rs747711475 Chr.17: 43191483-43191483 on Build GRCh38
rs745769330 Chr.17: 43191419-43191419 on Build GRCh38
rs201172896 Chr.17: 43191398-43191398 on Build GRCh38
rs753743690 Chr.17: 43191450-43191450 on Build GRCh38
rs202122812 Chr.17: 43191425-43191425 on Build GRCh38
rs974800773 Chr.17: 43191486-43191486 on Build GRCh38
rs771818220 Chr.17: 43191322-43191322 on Build GRCh38
rs1020392068 Chr.17: 43191353-43191353 on Build GRCh38
rs777516028 Chr.17: 43191485-43191485 on Build GRCh38
rs115552058 Chr.17: 43191443-43191443 on Build GRCh38
rs1027933017 Chr.17: 43191489-43191489 on Build GRCh38
rs368122136 Chr.17: 43191399-43191399 on Build GRCh38
rs780685398 Chr.17: 43191303-43191303 on Build GRCh38
rs754976385 Chr.17: 43191451-43191451 on Build GRCh38
rs776806589 Chr.17: 43191345-43191345 on Build GRCh38
rs964569208 Chr.17: 43191458-43191458 on Build GRCh38
rs770387760 Chr.17: 43191355-43191355 on Build GRCh38
rs1011181421 Chr.17: 43191351-43191351 on Build GRCh38

View Full Product Details