Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93252704-93252963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
UniGene
Hs.654856
Gene Chromosome Location
Chr.9: 93184156-93327581 on Build GRCh38
Transcripts
NM_001282394.1 NM_006648.3 XM_005252137.3 XM_005252140.2 XM_005252141.2 XM_005252144.2 XM_011518926.2 XM_011518927.2 XM_011518928.2 XM_011518929.2 XM_011518930.2 XM_011518931.2 XM_011518932.2 XM_011518935.2 XM_011518936.2 XM_011518937.2 XM_011518939.2 XM_011518940.2 XM_017015045.1 XM_017015046.1 XM_017015047.1 XM_017015048.1 XM_017015049.1 XM_017015050.1 XM_017015051.1 XM_017015052.1 XM_017015053.1 XM_017015054.1 XM_017015055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111233 c.1867A>G p.T623A Chr.9: 93252915-93252915 on Build GRCh38
4139669 c.697G>T p.G233C Chr.9: 93252909-93252909 on Build GRCh38
5159182 c.1883C>T p.S628L Chr.9: 93252931-93252931 on Build GRCh38
4139668 c.1861G>T p.G621C Chr.9: 93252909-93252909 on Build GRCh38
5691012 c.1836G>C p.S612S Chr.9: 93252884-93252884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533826158 Chr.9: 93252893-93252893 on Build GRCh38
rs555364985 Chr.9: 93252907-93252907 on Build GRCh38
rs865972199 Chr.9: 93252904-93252904 on Build GRCh38
rs201471342 Chr.9: 93252883-93252883 on Build GRCh38
rs777034860 Chr.9: 93252845-93252867 on Build GRCh38
rs764945379 Chr.9: 93252902-93252902 on Build GRCh38
rs553438989 Chr.9: 93252931-93252931 on Build GRCh38
rs756580132 Chr.9: 93252938-93252938 on Build GRCh38
rs967870922 Chr.9: 93252759-93252759 on Build GRCh38
rs991765250 Chr.9: 93252887-93252887 on Build GRCh38
rs776731494 Chr.9: 93252896-93252896 on Build GRCh38
rs969831947 Chr.9: 93252757-93252757 on Build GRCh38
rs778482022 Chr.9: 93252823-93252823 on Build GRCh38
rs762644931 Chr.9: 93252913-93252913 on Build GRCh38
rs780636320 Chr.9: 93252932-93252932 on Build GRCh38
rs774708109 Chr.9: 93252884-93252884 on Build GRCh38
rs928633319 Chr.9: 93252765-93252765 on Build GRCh38
rs756758043 Chr.9: 93252768-93252768 on Build GRCh38
rs759727110 Chr.9: 93252888-93252888 on Build GRCh38
rs1014123837 Chr.9: 93252728-93252728 on Build GRCh38
rs766123567 Chr.9: 93252915-93252915 on Build GRCh38
rs761801783 Chr.9: 93252900-93252900 on Build GRCh38
rs977396417 Chr.9: 93252763-93252763 on Build GRCh38
rs748952630 Chr.9: 93252753-93252753 on Build GRCh38
rs751183257 Chr.9: 93252918-93252918 on Build GRCh38
rs749725985 Chr.9: 93252942-93252942 on Build GRCh38
rs778015276 Chr.9: 93252940-93252940 on Build GRCh38
rs541692138 Chr.9: 93252853-93252853 on Build GRCh38
rs777315749 Chr.9: 93252738-93252738 on Build GRCh38

View Full Product Details