Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72132976-72133249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC1
Entrez Gene ID
51008
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
ASC1p50, CGI-18, SMABF2, p50
UniGene
Hs.500007
Gene Chromosome Location
Chr.10: 72096032-72217134 on Build GRCh38
Transcripts
NM_001198798.2 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016296.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1 XM_017016303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997029 c.846T>C p.V282V Chr.10: 72133166-72133166 on Build GRCh38
4924993 c.891A>G p.K297K Chr.10: 72133121-72133121 on Build GRCh38
3440323 c.766C>T p.R256* Chr.10: 72133162-72133162 on Build GRCh38
4602588 c.875A>G p.E292G Chr.10: 72133119-72133119 on Build GRCh38
3440322 c.850C>T p.R284* Chr.10: 72133162-72133162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748354891 Chr.10: 72133133-72133133 on Build GRCh38
rs779557354 Chr.10: 72133145-72133146 on Build GRCh38
rs113569227 Chr.10: 72133181-72133181 on Build GRCh38
rs752947666 Chr.10: 72133217-72133217 on Build GRCh38
rs773916542 Chr.10: 72133047-72133047 on Build GRCh38
rs758996240 Chr.10: 72133154-72133154 on Build GRCh38
rs771359121 Chr.10: 72133149-72133149 on Build GRCh38
rs377388692 Chr.10: 72133200-72133200 on Build GRCh38
rs756355497 Chr.10: 72133223-72133223 on Build GRCh38
rs142117888 Chr.10: 72133013-72133013 on Build GRCh38
rs773734112 Chr.10: 72133042-72133042 on Build GRCh38
rs1003697823 Chr.10: 72133159-72133159 on Build GRCh38
rs377549482 Chr.10: 72133017-72133017 on Build GRCh38
rs372834128 Chr.10: 72133043-72133043 on Build GRCh38
rs753803060 Chr.10: 72133058-72133058 on Build GRCh38
rs750332602 Chr.10: 72133096-72133097 on Build GRCh38
rs760488500 Chr.10: 72133053-72133053 on Build GRCh38
rs756007889 Chr.10: 72133142-72133143 on Build GRCh38
rs755134952 Chr.10: 72133099-72133099 on Build GRCh38
rs765897386 Chr.10: 72133046-72133046 on Build GRCh38
rs141611243 Chr.10: 72133162-72133162 on Build GRCh38
rs997267683 Chr.10: 72133060-72133060 on Build GRCh38
rs756286851 Chr.10: 72133222-72133223 on Build GRCh38
rs574845267 Chr.10: 72133049-72133049 on Build GRCh38
rs202208603 Chr.10: 72133041-72133041 on Build GRCh38
rs918992057 Chr.10: 72133220-72133220 on Build GRCh38
rs777009724 Chr.10: 72133026-72133026 on Build GRCh38
rs770170337 Chr.10: 72133134-72133134 on Build GRCh38
rs753511548 Chr.10: 72133187-72133187 on Build GRCh38
rs141996717 Chr.10: 72133185-72133185 on Build GRCh38
rs146370051 Chr.10: 72133059-72133059 on Build GRCh38
rs1011036993 Chr.10: 72133068-72133068 on Build GRCh38
rs565054047 Chr.10: 72133210-72133210 on Build GRCh38
rs778054296 Chr.10: 72133144-72133144 on Build GRCh38
rs751635146 Chr.10: 72133082-72133082 on Build GRCh38
rs768398422 Chr.10: 72133191-72133191 on Build GRCh38
rs943236533 Chr.10: 72133036-72133036 on Build GRCh38
rs376799409 Chr.10: 72133076-72133076 on Build GRCh38
rs371450934 Chr.10: 72133150-72133150 on Build GRCh38
rs1033847535 Chr.10: 72133203-72133203 on Build GRCh38
rs781528301 Chr.10: 72133116-72133116 on Build GRCh38
rs763845791 Chr.10: 72133056-72133056 on Build GRCh38
rs748499250 Chr.10: 72133030-72133030 on Build GRCh38
rs200721922 Chr.10: 72133088-72133088 on Build GRCh38
rs769419047 Chr.10: 72133016-72133016 on Build GRCh38
rs748854995 Chr.10: 72133147-72133148 on Build GRCh38
rs374955301 Chr.10: 72133050-72133050 on Build GRCh38

View Full Product Details